Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 65762941-65763509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT6P1
Entrez Gene ID
643253
Gene Name
chaperonin containing TCP1 subunit 6 pseudogene 1
Gene Aliases
CCT6-5P, CCT6AP1
UniGene
Hs.488399
Gene Chromosome Location
Chr.7: 65751105-65763675 on Build GRCh38
Transcripts
NR_003110.2 NR_038378.4

Gene Symbol
LOC441242
Entrez Gene ID
441242
Gene Name
uncharacterized LOC441242
Gene Aliases
-
UniGene
Hs.373941
Gene Chromosome Location
Chr.7: 65647010-65770810 on Build GRCh38
Transcripts
NR_003110.2 NR_038378.4

SNP Detail

SNP ID Chromosome Location
rs565613486 Chr.7: 65763142-65763142 on Build GRCh38
rs777701919 Chr.7: 65763021-65763021 on Build GRCh38
rs545471680 Chr.7: 65763374-65763374 on Build GRCh38
rs559991526 Chr.7: 65763338-65763338 on Build GRCh38
rs572093906 Chr.7: 65763093-65763093 on Build GRCh38
rs370478302 Chr.7: 65763126-65763126 on Build GRCh38
rs782569185 Chr.7: 65763068-65763069 on Build GRCh38
rs577557692 Chr.7: 65763230-65763230 on Build GRCh38
rs557667057 Chr.7: 65763261-65763261 on Build GRCh38
rs543214514 Chr.7: 65763315-65763315 on Build GRCh38
rs535026108 Chr.7: 65763281-65763281 on Build GRCh38
rs528535968 Chr.7: 65763365-65763365 on Build GRCh38
rs535254360 Chr.7: 65762989-65762989 on Build GRCh38
rs565279897 Chr.7: 65763400-65763400 on Build GRCh38
rs543490802 Chr.7: 65763257-65763257 on Build GRCh38
rs574422022 Chr.7: 65763301-65763301 on Build GRCh38
rs71045099 Chr.7: 65763332-65763333 on Build GRCh38
rs534722431 Chr.7: 65763196-65763196 on Build GRCh38
rs555161417 Chr.7: 65763012-65763012 on Build GRCh38
rs368094433 Chr.7: 65763276-65763276 on Build GRCh38
rs557431282 Chr.7: 65763211-65763211 on Build GRCh38

View Full Product Details