Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 92575581-92576128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM133B
Entrez Gene ID
257415
Gene Name
family with sequence similarity 133 member B
Gene Aliases
-
UniGene
Hs.18564
Gene Chromosome Location
Chr.7: 92560758-92590394 on Build GRCh38
Transcripts
NM_001040057.2 NM_001288584.1 NM_152789.3 NR_109929.1 XM_011516004.2 XM_011516005.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3663405 c.516+4A>C p.? Chr.7: 92575767-92575767 on Build GRCh38
3431864 c.486G>T p.K162N Chr.7: 92575801-92575801 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs17760432 Chr.7: 92575837-92575837 on Build GRCh38
rs7794389 Chr.7: 92576044-92576044 on Build GRCh38
rs553152117 Chr.7: 92576072-92576072 on Build GRCh38
rs1055490973 Chr.7: 92575919-92575919 on Build GRCh38
rs755041869 Chr.7: 92575733-92575733 on Build GRCh38
rs10250762 Chr.7: 92576038-92576038 on Build GRCh38
rs990959352 Chr.7: 92576103-92576103 on Build GRCh38
rs771265660 Chr.7: 92575790-92575790 on Build GRCh38
rs956985216 Chr.7: 92575788-92575788 on Build GRCh38
rs750398425 Chr.7: 92575724-92575724 on Build GRCh38
rs996666258 Chr.7: 92575993-92575993 on Build GRCh38
rs115236001 Chr.7: 92575681-92575681 on Build GRCh38
rs187233489 Chr.7: 92575749-92575749 on Build GRCh38
rs576214815 Chr.7: 92576098-92576098 on Build GRCh38
rs771642506 Chr.7: 92575820-92575820 on Build GRCh38
rs541048697 Chr.7: 92575925-92575925 on Build GRCh38
rs1052424808 Chr.7: 92575991-92575991 on Build GRCh38
rs979886739 Chr.7: 92575839-92575839 on Build GRCh38
rs199977160 Chr.7: 92575740-92575740 on Build GRCh38
rs936677077 Chr.7: 92575978-92575978 on Build GRCh38
rs192856610 Chr.7: 92575846-92575846 on Build GRCh38
rs949774872 Chr.7: 92575933-92575933 on Build GRCh38
rs926931338 Chr.7: 92575920-92575920 on Build GRCh38
rs199719983 Chr.7: 92575626-92575626 on Build GRCh38
rs1017258478 Chr.7: 92575965-92575965 on Build GRCh38
rs945667128 Chr.7: 92575880-92575880 on Build GRCh38
rs775172144 Chr.7: 92575836-92575836 on Build GRCh38
rs925508513 Chr.7: 92575913-92575913 on Build GRCh38
rs892700095 Chr.7: 92576019-92576019 on Build GRCh38
rs967105545 Chr.7: 92576055-92576055 on Build GRCh38
rs777904335 Chr.7: 92575767-92575767 on Build GRCh38
rs756979439 Chr.7: 92575952-92575952 on Build GRCh38
rs866255343 Chr.7: 92576096-92576096 on Build GRCh38
rs985080772 Chr.7: 92575943-92575943 on Build GRCh38
rs752872789 Chr.7: 92575743-92575743 on Build GRCh38
rs938175223 Chr.7: 92575917-92575917 on Build GRCh38
rs898369838 Chr.7: 92575975-92575975 on Build GRCh38
rs761666209 Chr.7: 92575866-92575866 on Build GRCh38
rs1028409860 Chr.7: 92575751-92575751 on Build GRCh38
rs74635435 Chr.7: 92575953-92575953 on Build GRCh38
rs1001654604 Chr.7: 92576057-92576057 on Build GRCh38
rs980054230 Chr.7: 92575886-92575886 on Build GRCh38
rs539191098 Chr.7: 92575646-92575646 on Build GRCh38
rs746391073 Chr.7: 92575813-92575813 on Build GRCh38
rs533826090 Chr.7: 92575636-92575636 on Build GRCh38
rs75520111 Chr.7: 92576095-92576095 on Build GRCh38
rs776188719 Chr.7: 92575861-92575861 on Build GRCh38
rs564391165 Chr.7: 92575944-92575944 on Build GRCh38
rs1016767989 Chr.7: 92575731-92575731 on Build GRCh38
rs866529757 Chr.7: 92575937-92575937 on Build GRCh38
rs766634445 Chr.7: 92576026-92576026 on Build GRCh38
rs749631536 Chr.7: 92575773-92575773 on Build GRCh38
rs979747304 Chr.7: 92576056-92576056 on Build GRCh38
rs1050536072 Chr.7: 92575971-92575971 on Build GRCh38
rs1040267277 Chr.7: 92575673-92575673 on Build GRCh38
rs781725567 Chr.7: 92576069-92576069 on Build GRCh38
rs903818442 Chr.7: 92575972-92575973 on Build GRCh38
rs372774049 Chr.7: 92575912-92575912 on Build GRCh38
rs996300232 Chr.7: 92575720-92575720 on Build GRCh38
rs748892198 Chr.7: 92575770-92575770 on Build GRCh38
rs576896302 Chr.7: 92575618-92575618 on Build GRCh38
rs531579064 Chr.7: 92576046-92576046 on Build GRCh38
rs779461130 Chr.7: 92575806-92575806 on Build GRCh38
rs984472685 Chr.7: 92576091-92576091 on Build GRCh38
rs576605016 Chr.7: 92575696-92575696 on Build GRCh38

View Full Product Details