Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 103174808-103175362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPY19L2P2
Entrez Gene ID
349152
Gene Name
DPY19L2 pseudogene 2
Gene Aliases
-
UniGene
Hs.732579
Gene Chromosome Location
Chr.7: 103175013-103280466 on Build GRCh38
Transcripts
NR_003561.2 NR_027768.1

SNP Detail

SNP ID Chromosome Location
rs890493778 Chr.7: 103175112-103175112 on Build GRCh38
rs556698229 Chr.7: 103175012-103175012 on Build GRCh38
rs951253696 Chr.7: 103174943-103174943 on Build GRCh38
rs535065297 Chr.7: 103174993-103174993 on Build GRCh38
rs2411096 Chr.7: 103175194-103175194 on Build GRCh38
rs553884436 Chr.7: 103175058-103175058 on Build GRCh38
rs575270918 Chr.7: 103175037-103175037 on Build GRCh38
rs951181752 Chr.7: 103175195-103175195 on Build GRCh38
rs776787504 Chr.7: 103175254-103175255 on Build GRCh38
rs926386261 Chr.7: 103174981-103174982 on Build GRCh38
rs781466655 Chr.7: 103175050-103175051 on Build GRCh38
rs2411095 Chr.7: 103175148-103175148 on Build GRCh38
rs3987920 Chr.7: 103175105-103175105 on Build GRCh38
rs995764644 Chr.7: 103175169-103175169 on Build GRCh38
rs1053921066 Chr.7: 103175036-103175036 on Build GRCh38
rs1024895571 Chr.7: 103175211-103175211 on Build GRCh38
rs1000863787 Chr.7: 103175026-103175026 on Build GRCh38
rs74788641 Chr.7: 103175272-103175272 on Build GRCh38
rs970363356 Chr.7: 103174971-103174971 on Build GRCh38
rs957790438 Chr.7: 103175270-103175271 on Build GRCh38
rs530452142 Chr.7: 103175295-103175295 on Build GRCh38
rs138089416 Chr.7: 103175147-103175148 on Build GRCh38
rs970835465 Chr.7: 103175237-103175237 on Build GRCh38
rs2909184 Chr.7: 103174973-103174973 on Build GRCh38
rs1026789834 Chr.7: 103175175-103175175 on Build GRCh38
rs3987921 Chr.7: 103175111-103175111 on Build GRCh38
rs980431748 Chr.7: 103175259-103175259 on Build GRCh38
rs577505954 Chr.7: 103175262-103175262 on Build GRCh38
rs560066391 Chr.7: 103175278-103175278 on Build GRCh38
rs1007548785 Chr.7: 103175145-103175145 on Build GRCh38
rs899208940 Chr.7: 103175148-103175149 on Build GRCh38
rs188608215 Chr.7: 103175268-103175268 on Build GRCh38
rs2411094 Chr.7: 103175119-103175119 on Build GRCh38
rs776721854 Chr.7: 103175253-103175254 on Build GRCh38
rs750230970 Chr.7: 103175257-103175258 on Build GRCh38
rs376180616 Chr.7: 103175260-103175260 on Build GRCh38
rs148045205 Chr.7: 103175063-103175063 on Build GRCh38
rs62480095 Chr.7: 103175274-103175274 on Build GRCh38
rs568039379 Chr.7: 103174964-103174964 on Build GRCh38
rs397842197 Chr.7: 103174972-103174972 on Build GRCh38
rs939897327 Chr.7: 103175332-103175332 on Build GRCh38
rs7796853 Chr.7: 103175258-103175258 on Build GRCh38
rs762806720 Chr.7: 103175254-103175254 on Build GRCh38
rs758618653 Chr.7: 103175258-103175258 on Build GRCh38
rs1004157108 Chr.7: 103175204-103175204 on Build GRCh38
rs982457362 Chr.7: 103174953-103174953 on Build GRCh38
rs936844369 Chr.7: 103175011-103175011 on Build GRCh38
rs3987919 Chr.7: 103175010-103175010 on Build GRCh38
rs765421309 Chr.7: 103175258-103175259 on Build GRCh38
rs3959196 Chr.7: 103175089-103175089 on Build GRCh38
rs370014298 Chr.7: 103175148-103175149 on Build GRCh38

View Full Product Details