Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 103184068-103184615 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPY19L2P2
Entrez Gene ID
349152
Gene Name
DPY19L2 pseudogene 2
Gene Aliases
-
UniGene
Hs.732579
Gene Chromosome Location
Chr.7: 103175013-103280466 on Build GRCh38
Transcripts
NR_003561.2 NR_027768.1

SNP Detail

SNP ID Chromosome Location
rs191609910 Chr.7: 103184529-103184529 on Build GRCh38
rs541825972 Chr.7: 103184230-103184230 on Build GRCh38
rs552632076 Chr.7: 103184167-103184169 on Build GRCh38
rs923583351 Chr.7: 103184242-103184242 on Build GRCh38
rs1034360217 Chr.7: 103184409-103184410 on Build GRCh38
rs894992743 Chr.7: 103184417-103184417 on Build GRCh38
rs968133970 Chr.7: 103184196-103184196 on Build GRCh38
rs138373846 Chr.7: 103184470-103184470 on Build GRCh38
rs528247642 Chr.7: 103184477-103184477 on Build GRCh38
rs932098762 Chr.7: 103184285-103184285 on Build GRCh38
rs4301376 Chr.7: 103184205-103184205 on Build GRCh38
rs965696845 Chr.7: 103184560-103184560 on Build GRCh38
rs888006452 Chr.7: 103184295-103184295 on Build GRCh38
rs372300799 Chr.7: 103184426-103184426 on Build GRCh38
rs751830615 Chr.7: 103184293-103184293 on Build GRCh38
rs915130834 Chr.7: 103184194-103184194 on Build GRCh38
rs553369667 Chr.7: 103184146-103184146 on Build GRCh38
rs76551380 Chr.7: 103184445-103184445 on Build GRCh38
rs1036395285 Chr.7: 103184354-103184354 on Build GRCh38
rs975831950 Chr.7: 103184578-103184578 on Build GRCh38
rs959196890 Chr.7: 103184144-103184144 on Build GRCh38
rs907190376 Chr.7: 103184406-103184406 on Build GRCh38
rs922132197 Chr.7: 103184257-103184257 on Build GRCh38
rs940705158 Chr.7: 103184345-103184345 on Build GRCh38
rs1040137014 Chr.7: 103184256-103184256 on Build GRCh38
rs185912097 Chr.7: 103184407-103184407 on Build GRCh38
rs1012007335 Chr.7: 103184440-103184440 on Build GRCh38
rs1030698446 Chr.7: 103184525-103184525 on Build GRCh38
rs4330612 Chr.7: 103184468-103184468 on Build GRCh38
rs944466406 Chr.7: 103184255-103184255 on Build GRCh38
rs1034851064 Chr.7: 103184128-103184128 on Build GRCh38

View Full Product Details