Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 103210283-103210697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPY19L2P2
Entrez Gene ID
349152
Gene Name
DPY19L2 pseudogene 2
Gene Aliases
-
UniGene
Hs.732579
Gene Chromosome Location
Chr.7: 103175013-103280466 on Build GRCh38
Transcripts
NR_003561.2 NR_027768.1

SNP Detail

SNP ID Chromosome Location
rs772334594 Chr.7: 103210636-103210645 on Build GRCh38
rs972526644 Chr.7: 103210531-103210531 on Build GRCh38
rs144177747 Chr.7: 103210630-103210641 on Build GRCh38
rs202062307 Chr.7: 103210630-103210639 on Build GRCh38
rs903966852 Chr.7: 103210350-103210350 on Build GRCh38
rs561265552 Chr.7: 103210520-103210520 on Build GRCh38
rs772061202 Chr.7: 103210655-103210671 on Build GRCh38
rs145022643 Chr.7: 103210665-103210665 on Build GRCh38
rs368364671 Chr.7: 103210671-103210672 on Build GRCh38
rs997983124 Chr.7: 103210426-103210426 on Build GRCh38
rs756659146 Chr.7: 103210318-103210318 on Build GRCh38
rs11560368 Chr.7: 103210547-103210547 on Build GRCh38
rs9886017 Chr.7: 103210671-103210671 on Build GRCh38
rs145456276 Chr.7: 103210630-103210645 on Build GRCh38
rs181038848 Chr.7: 103210502-103210502 on Build GRCh38
rs1031482377 Chr.7: 103210397-103210397 on Build GRCh38
rs369789520 Chr.7: 103210647-103210648 on Build GRCh38
rs867905175 Chr.7: 103210655-103210655 on Build GRCh38
rs62481612 Chr.7: 103210484-103210484 on Build GRCh38
rs916864428 Chr.7: 103210600-103210600 on Build GRCh38
rs554191102 Chr.7: 103210469-103210469 on Build GRCh38
rs764754983 Chr.7: 103210321-103210321 on Build GRCh38
rs556846796 Chr.7: 103210365-103210365 on Build GRCh38
rs538066823 Chr.7: 103210339-103210339 on Build GRCh38
rs758694277 Chr.7: 103210359-103210359 on Build GRCh38
rs578254782 Chr.7: 103210374-103210374 on Build GRCh38
rs757564828 Chr.7: 103210326-103210326 on Build GRCh38
rs985564319 Chr.7: 103210497-103210497 on Build GRCh38
rs147974399 Chr.7: 103210667-103210667 on Build GRCh38
rs772746564 Chr.7: 103210307-103210307 on Build GRCh38
rs751164389 Chr.7: 103210311-103210311 on Build GRCh38
rs867250453 Chr.7: 103210651-103210651 on Build GRCh38
rs992284121 Chr.7: 103210579-103210579 on Build GRCh38
rs17136079 Chr.7: 103210546-103210546 on Build GRCh38
rs778493603 Chr.7: 103210632-103210645 on Build GRCh38
rs779155331 Chr.7: 103210336-103210336 on Build GRCh38
rs948320041 Chr.7: 103210631-103210631 on Build GRCh38
rs146108365 Chr.7: 103210669-103210669 on Build GRCh38
rs538998623 Chr.7: 103210464-103210464 on Build GRCh38
rs564802859 Chr.7: 103210619-103210619 on Build GRCh38

View Full Product Details