Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 103254565-103255135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPY19L2P2
Entrez Gene ID
349152
Gene Name
DPY19L2 pseudogene 2
Gene Aliases
-
UniGene
Hs.732579
Gene Chromosome Location
Chr.7: 103175013-103280466 on Build GRCh38
Transcripts
NR_003561.2 NR_027768.1

SNP Detail

SNP ID Chromosome Location
rs189750719 Chr.7: 103254928-103254928 on Build GRCh38
rs956868490 Chr.7: 103254923-103254923 on Build GRCh38
rs767616070 Chr.7: 103254869-103254869 on Build GRCh38
rs1038190418 Chr.7: 103254777-103254777 on Build GRCh38
rs529945175 Chr.7: 103255070-103255073 on Build GRCh38
rs184858074 Chr.7: 103254831-103254831 on Build GRCh38
rs748656383 Chr.7: 103254835-103254835 on Build GRCh38
rs759490012 Chr.7: 103254865-103254865 on Build GRCh38
rs181857410 Chr.7: 103255073-103255073 on Build GRCh38
rs1047430881 Chr.7: 103255035-103255036 on Build GRCh38
rs1046950580 Chr.7: 103254800-103254800 on Build GRCh38
rs745510938 Chr.7: 103254914-103254914 on Build GRCh38
rs745326538 Chr.7: 103254828-103254828 on Build GRCh38
rs145875184 Chr.7: 103254945-103254945 on Build GRCh38
rs896390902 Chr.7: 103255038-103255038 on Build GRCh38
rs933314064 Chr.7: 103254652-103254652 on Build GRCh38
rs1037759683 Chr.7: 103254995-103254995 on Build GRCh38
rs544929171 Chr.7: 103254811-103254811 on Build GRCh38
rs752702748 Chr.7: 103254812-103254812 on Build GRCh38
rs778377037 Chr.7: 103254902-103254902 on Build GRCh38
rs7796589 Chr.7: 103254819-103254819 on Build GRCh38
rs145333746 Chr.7: 103255078-103255081 on Build GRCh38
rs1009707258 Chr.7: 103255078-103255079 on Build GRCh38
rs764137700 Chr.7: 103254876-103254876 on Build GRCh38
rs544860335 Chr.7: 103255094-103255094 on Build GRCh38
rs747176223 Chr.7: 103254735-103254735 on Build GRCh38
rs562314910 Chr.7: 103254787-103254787 on Build GRCh38
rs767318064 Chr.7: 103254810-103254810 on Build GRCh38
rs758747508 Chr.7: 103255037-103255038 on Build GRCh38
rs775855259 Chr.7: 103255071-103255071 on Build GRCh38
rs904983880 Chr.7: 103255067-103255068 on Build GRCh38
rs766521691 Chr.7: 103254849-103254849 on Build GRCh38
rs1050946417 Chr.7: 103254719-103254719 on Build GRCh38
rs572167863 Chr.7: 103255067-103255067 on Build GRCh38
rs7777767 Chr.7: 103254756-103254756 on Build GRCh38
rs777226770 Chr.7: 103254759-103254759 on Build GRCh38
rs538698410 Chr.7: 103254599-103254599 on Build GRCh38
rs551778681 Chr.7: 103254829-103254829 on Build GRCh38
rs574335989 Chr.7: 103254796-103254796 on Build GRCh38
rs765561237 Chr.7: 103254785-103254785 on Build GRCh38
rs562865034 Chr.7: 103254817-103254817 on Build GRCh38
rs757145325 Chr.7: 103254901-103254901 on Build GRCh38
rs142458825 Chr.7: 103255036-103255037 on Build GRCh38
rs770351241 Chr.7: 103254840-103254840 on Build GRCh38
rs189506709 Chr.7: 103255092-103255092 on Build GRCh38
rs752763162 Chr.7: 103254870-103254870 on Build GRCh38
rs756170759 Chr.7: 103254816-103254816 on Build GRCh38
rs527830310 Chr.7: 103254898-103254898 on Build GRCh38
rs773982865 Chr.7: 103254733-103254733 on Build GRCh38
rs756910673 Chr.7: 103254826-103254826 on Build GRCh38
rs548722663 Chr.7: 103254919-103254919 on Build GRCh38
rs773554885 Chr.7: 103254844-103254844 on Build GRCh38
rs567135045 Chr.7: 103254926-103254926 on Build GRCh38
rs954984621 Chr.7: 103255079-103255079 on Build GRCh38
rs751539860 Chr.7: 103254791-103254791 on Build GRCh38
rs762030538 Chr.7: 103254770-103254770 on Build GRCh38
rs778631232 Chr.7: 103254827-103254827 on Build GRCh38
rs7781624 Chr.7: 103255004-103255004 on Build GRCh38
rs923510217 Chr.7: 103254942-103254942 on Build GRCh38
rs923279670 Chr.7: 103254610-103254610 on Build GRCh38
rs553770041 Chr.7: 103255055-103255055 on Build GRCh38
rs988819274 Chr.7: 103254939-103254939 on Build GRCh38
rs540820365 Chr.7: 103254786-103254786 on Build GRCh38
rs745394971 Chr.7: 103254741-103254741 on Build GRCh38
rs184296085 Chr.7: 103255080-103255080 on Build GRCh38
rs763322235 Chr.7: 103254845-103254845 on Build GRCh38
rs553565578 Chr.7: 103254640-103254640 on Build GRCh38
rs774664526 Chr.7: 103254850-103254850 on Build GRCh38
rs770052566 Chr.7: 103254965-103254965 on Build GRCh38
rs571547714 Chr.7: 103254958-103254958 on Build GRCh38
rs755168486 Chr.7: 103255061-103255062 on Build GRCh38
rs910629487 Chr.7: 103254726-103254726 on Build GRCh38
rs1000576981 Chr.7: 103255068-103255068 on Build GRCh38
rs929786128 Chr.7: 103255016-103255016 on Build GRCh38

View Full Product Details