Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 103278193-103278785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPY19L2P2
Entrez Gene ID
349152
Gene Name
DPY19L2 pseudogene 2
Gene Aliases
-
UniGene
Hs.732579
Gene Chromosome Location
Chr.7: 103175013-103280466 on Build GRCh38
Transcripts
NR_003561.2 NR_027768.1

SNP Detail

SNP ID Chromosome Location
rs540749702 Chr.7: 103278748-103278748 on Build GRCh38
rs746596706 Chr.7: 103278271-103278271 on Build GRCh38
rs185637937 Chr.7: 103278522-103278522 on Build GRCh38
rs193039954 Chr.7: 103278341-103278341 on Build GRCh38
rs1030617894 Chr.7: 103278637-103278637 on Build GRCh38
rs954559205 Chr.7: 103278658-103278658 on Build GRCh38
rs537846318 Chr.7: 103278275-103278275 on Build GRCh38
rs373498780 Chr.7: 103278237-103278237 on Build GRCh38
rs113979352 Chr.7: 103278641-103278641 on Build GRCh38
rs998261330 Chr.7: 103278621-103278621 on Build GRCh38
rs143250949 Chr.7: 103278566-103278566 on Build GRCh38
rs377492323 Chr.7: 103278402-103278402 on Build GRCh38
rs933254732 Chr.7: 103278370-103278370 on Build GRCh38
rs1013156081 Chr.7: 103278223-103278223 on Build GRCh38
rs372386867 Chr.7: 103278449-103278449 on Build GRCh38
rs371087408 Chr.7: 103278687-103278687 on Build GRCh38
rs923115353 Chr.7: 103278369-103278369 on Build GRCh38
rs778497040 Chr.7: 103278245-103278245 on Build GRCh38
rs546496299 Chr.7: 103278291-103278291 on Build GRCh38
rs576845899 Chr.7: 103278734-103278734 on Build GRCh38
rs1013627099 Chr.7: 103278534-103278534 on Build GRCh38
rs748220830 Chr.7: 103278254-103278254 on Build GRCh38
rs188966036 Chr.7: 103278647-103278647 on Build GRCh38
rs751939229 Chr.7: 103278312-103278313 on Build GRCh38
rs1023167639 Chr.7: 103278551-103278551 on Build GRCh38
rs542882198 Chr.7: 103278517-103278517 on Build GRCh38
rs556227180 Chr.7: 103278699-103278702 on Build GRCh38
rs567659766 Chr.7: 103278258-103278258 on Build GRCh38
rs554268342 Chr.7: 103278439-103278439 on Build GRCh38
rs116544918 Chr.7: 103278474-103278474 on Build GRCh38
rs986043264 Chr.7: 103278386-103278386 on Build GRCh38
rs988390341 Chr.7: 103278367-103278367 on Build GRCh38
rs543405894 Chr.7: 103278599-103278599 on Build GRCh38
rs781571043 Chr.7: 103278221-103278221 on Build GRCh38
rs910252825 Chr.7: 103278409-103278409 on Build GRCh38
rs770925744 Chr.7: 103278309-103278309 on Build GRCh38
rs1037331447 Chr.7: 103278452-103278452 on Build GRCh38
rs774122561 Chr.7: 103278343-103278343 on Build GRCh38
rs956346356 Chr.7: 103278609-103278609 on Build GRCh38
rs538778300 Chr.7: 103278392-103278392 on Build GRCh38
rs10249842 Chr.7: 103278308-103278308 on Build GRCh38
rs868465799 Chr.7: 103278600-103278600 on Build GRCh38
rs985927041 Chr.7: 103278681-103278681 on Build GRCh38
rs910474038 Chr.7: 103278702-103278702 on Build GRCh38
rs895969289 Chr.7: 103278528-103278528 on Build GRCh38
rs904752857 Chr.7: 103278553-103278553 on Build GRCh38
rs941684282 Chr.7: 103278428-103278428 on Build GRCh38

View Full Product Details