Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1486434-1486783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATAD3B
Entrez Gene ID
83858
Gene Name
ATPase family, AAA domain containing 3B
Gene Aliases
AAA-TOB3, TOB3
UniGene
Hs.729021
Gene Chromosome Location
Chr.1: 1471732-1509466 on Build GRCh38
Transcripts
NM_001317238.1 NM_031921.5 XM_005244806.3 XM_011542241.2 XM_011542243.2 XM_011542244.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2230222 c.1111A>T p.M371L Chr.1: 1486565-1486565 on Build GRCh38
5003811 c.1120G>A p.A374T Chr.1: 1486574-1486574 on Build GRCh38
3747530 c.1157G>A p.R386Q Chr.1: 1486611-1486611 on Build GRCh38
4663944 c.1142delT p.V381fs*9 Chr.1: 1486596-1486596 on Build GRCh38
6120713 c.1118A>T p.Y373F Chr.1: 1486572-1486572 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770117658 Chr.1: 1486662-1486662 on Build GRCh38
rs777740137 Chr.1: 1486506-1486506 on Build GRCh38
rs753559374 Chr.1: 1486624-1486624 on Build GRCh38
rs879237985 Chr.1: 1486473-1486473 on Build GRCh38
rs1013173944 Chr.1: 1486453-1486453 on Build GRCh38
rs767884119 Chr.1: 1486495-1486495 on Build GRCh38
rs752516554 Chr.1: 1486500-1486501 on Build GRCh38
rs922481686 Chr.1: 1486601-1486601 on Build GRCh38
rs774752691 Chr.1: 1486517-1486517 on Build GRCh38
rs745516549 Chr.1: 1486510-1486510 on Build GRCh38
rs994045943 Chr.1: 1486530-1486530 on Build GRCh38
rs747479353 Chr.1: 1486650-1486650 on Build GRCh38
rs371036662 Chr.1: 1486482-1486482 on Build GRCh38
rs758401413 Chr.1: 1486613-1486613 on Build GRCh38
rs993747880 Chr.1: 1486749-1486749 on Build GRCh38
rs137911966 Chr.1: 1486574-1486574 on Build GRCh38
rs773854999 Chr.1: 1486582-1486582 on Build GRCh38
rs572142577 Chr.1: 1486594-1486594 on Build GRCh38
rs759684788 Chr.1: 1486610-1486610 on Build GRCh38
rs760632402 Chr.1: 1486670-1486670 on Build GRCh38
rs779902355 Chr.1: 1486572-1486572 on Build GRCh38
rs768257981 Chr.1: 1486515-1486515 on Build GRCh38
rs570011887 Chr.1: 1486564-1486564 on Build GRCh38
rs750391230 Chr.1: 1486667-1486667 on Build GRCh38
rs542996639 Chr.1: 1486461-1486461 on Build GRCh38
rs780244099 Chr.1: 1486500-1486500 on Build GRCh38
rs576024748 Chr.1: 1486651-1486651 on Build GRCh38
rs371473559 Chr.1: 1486757-1486757 on Build GRCh38
rs746912693 Chr.1: 1486507-1486507 on Build GRCh38
rs148189584 Chr.1: 1486584-1486584 on Build GRCh38
rs754700272 Chr.1: 1486689-1486689 on Build GRCh38
rs757029683 Chr.1: 1486508-1486508 on Build GRCh38
rs201951488 Chr.1: 1486536-1486536 on Build GRCh38
rs750616633 Chr.1: 1486497-1486497 on Build GRCh38
rs955304836 Chr.1: 1486605-1486605 on Build GRCh38
rs141377718 Chr.1: 1486595-1486595 on Build GRCh38
rs778680032 Chr.1: 1486696-1486696 on Build GRCh38
rs771481965 Chr.1: 1486709-1486709 on Build GRCh38
rs769627263 Chr.1: 1486513-1486513 on Build GRCh38
rs780631900 Chr.1: 1486642-1486645 on Build GRCh38
rs860213 Chr.1: 1486611-1486611 on Build GRCh38
rs758105453 Chr.1: 1486523-1486524 on Build GRCh38
rs776918976 Chr.1: 1486654-1486654 on Build GRCh38
rs1046362931 Chr.1: 1486481-1486481 on Build GRCh38
rs547746106 Chr.1: 1486454-1486454 on Build GRCh38
rs367883442 Chr.1: 1486707-1486707 on Build GRCh38
rs757157792 Chr.1: 1486550-1486550 on Build GRCh38
rs756303826 Chr.1: 1486498-1486498 on Build GRCh38
rs139682140 Chr.1: 1486661-1486661 on Build GRCh38
rs751073167 Chr.1: 1486531-1486533 on Build GRCh38
rs778369695 Chr.1: 1486639-1486639 on Build GRCh38
rs371563971 Chr.1: 1486708-1486708 on Build GRCh38
rs776865693 Chr.1: 1486604-1486604 on Build GRCh38
rs779928923 Chr.1: 1486620-1486620 on Build GRCh38
rs766407596 Chr.1: 1486675-1486675 on Build GRCh38
rs754040873 Chr.1: 1486534-1486534 on Build GRCh38
rs777399500 Chr.1: 1486526-1486529 on Build GRCh38
rs768286643 Chr.1: 1486575-1486575 on Build GRCh38
rs907480205 Chr.1: 1486733-1486733 on Build GRCh38
rs557549921 Chr.1: 1486619-1486619 on Build GRCh38
rs200307284 Chr.1: 1486526-1486526 on Build GRCh38
rs140644969 Chr.1: 1486565-1486565 on Build GRCh38
rs201713686 Chr.1: 1486665-1486665 on Build GRCh38
rs752073214 Chr.1: 1486493-1486493 on Build GRCh38
rs752246226 Chr.1: 1486697-1486697 on Build GRCh38
rs772706411 Chr.1: 1486588-1486588 on Build GRCh38
rs762315264 Chr.1: 1486521-1486521 on Build GRCh38
rs754737173 Chr.1: 1486625-1486625 on Build GRCh38
rs541977955 Chr.1: 1486545-1486545 on Build GRCh38
rs756697728 Chr.1: 1486621-1486622 on Build GRCh38
rs767788401 Chr.1: 1486523-1486523 on Build GRCh38
rs964647958 Chr.1: 1486586-1486586 on Build GRCh38
rs780971076 Chr.1: 1486509-1486509 on Build GRCh38
rs374902984 Chr.1: 1486544-1486544 on Build GRCh38
rs755023603 Chr.1: 1486504-1486504 on Build GRCh38
rs771285533 Chr.1: 1486652-1486652 on Build GRCh38
rs558078851 Chr.1: 1486602-1486602 on Build GRCh38
rs543613252 Chr.1: 1486618-1486618 on Build GRCh38
rs539299884 Chr.1: 1486501-1486501 on Build GRCh38
rs771463932 Chr.1: 1486634-1486634 on Build GRCh38
rs368162702 Chr.1: 1486666-1486666 on Build GRCh38
rs184670322 Chr.1: 1486514-1486514 on Build GRCh38
rs780974870 Chr.1: 1486555-1486555 on Build GRCh38
rs1045996404 Chr.1: 1486733-1486733 on Build GRCh38
rs761025941 Chr.1: 1486529-1486529 on Build GRCh38
rs527322901 Chr.1: 1486678-1486678 on Build GRCh38
rs775542155 Chr.1: 1486664-1486664 on Build GRCh38
rs747736228 Chr.1: 1486633-1486633 on Build GRCh38

View Full Product Details