Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 106094454-106094850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AASDHPPT
Entrez Gene ID
60496
Gene Name
aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase
Gene Aliases
AASD-PPT, ACPS, CGI-80, LYS2, LYS5
UniGene
Hs.524009
Gene Chromosome Location
Chr.11: 106077565-106098692 on Build GRCh38
Transcripts
NM_015423.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6224571 c.744C>T p.P248P Chr.11: 106094633-106094633 on Build GRCh38
4168762 c.698G>A p.S233N Chr.11: 106094587-106094587 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs546175982 Chr.11: 106094634-106094634 on Build GRCh38
rs945353225 Chr.11: 106094737-106094737 on Build GRCh38
rs763849714 Chr.11: 106094681-106094681 on Build GRCh38
rs900432871 Chr.11: 106094781-106094781 on Build GRCh38
rs989488307 Chr.11: 106094668-106094668 on Build GRCh38
rs1893175 Chr.11: 106094489-106094489 on Build GRCh38
rs758251465 Chr.11: 106094702-106094702 on Build GRCh38
rs745749381 Chr.11: 106094569-106094569 on Build GRCh38
rs140111373 Chr.11: 106094732-106094732 on Build GRCh38
rs760165467 Chr.11: 106094672-106094672 on Build GRCh38
rs72977065 Chr.11: 106094487-106094487 on Build GRCh38
rs964864691 Chr.11: 106094483-106094483 on Build GRCh38
rs1042805729 Chr.11: 106094750-106094750 on Build GRCh38
rs1027472003 Chr.11: 106094811-106094811 on Build GRCh38
rs749098652 Chr.11: 106094545-106094545 on Build GRCh38
rs533003962 Chr.11: 106094478-106094478 on Build GRCh38
rs934391412 Chr.11: 106094637-106094637 on Build GRCh38
rs112187654 Chr.11: 106094739-106094739 on Build GRCh38
rs779014124 Chr.11: 106094567-106094567 on Build GRCh38
rs760227539 Chr.11: 106094710-106094710 on Build GRCh38
rs113357961 Chr.11: 106094491-106094491 on Build GRCh38
rs995193496 Chr.11: 106094785-106094785 on Build GRCh38
rs908544095 Chr.11: 106094517-106094517 on Build GRCh38
rs555375415 Chr.11: 106094476-106094477 on Build GRCh38
rs575819868 Chr.11: 106094553-106094553 on Build GRCh38
rs747326449 Chr.11: 106094608-106094608 on Build GRCh38
rs766040559 Chr.11: 106094743-106094743 on Build GRCh38
rs760559805 Chr.11: 106094669-106094669 on Build GRCh38
rs994550963 Chr.11: 106094520-106094520 on Build GRCh38
rs750420256 Chr.11: 106094698-106094698 on Build GRCh38
rs778817635 Chr.11: 106094690-106094690 on Build GRCh38
rs56149260 Chr.11: 106094674-106094674 on Build GRCh38
rs776779250 Chr.11: 106094625-106094625 on Build GRCh38
rs376752869 Chr.11: 106094783-106094783 on Build GRCh38
rs912133448 Chr.11: 106094757-106094758 on Build GRCh38
rs201664850 Chr.11: 106094688-106094688 on Build GRCh38
rs567915575 Chr.11: 106094478-106094479 on Build GRCh38
rs1027414606 Chr.11: 106094524-106094524 on Build GRCh38
rs752143900 Chr.11: 106094654-106094654 on Build GRCh38
rs916168107 Chr.11: 106094611-106094611 on Build GRCh38
rs768875145 Chr.11: 106094622-106094622 on Build GRCh38
rs973261920 Chr.11: 106094485-106094485 on Build GRCh38
rs377756672 Chr.11: 106094543-106094543 on Build GRCh38
rs201593324 Chr.11: 106094477-106094478 on Build GRCh38
rs762191204 Chr.11: 106094633-106094633 on Build GRCh38
rs775407217 Chr.11: 106094605-106094605 on Build GRCh38
rs773811849 Chr.11: 106094636-106094636 on Build GRCh38
rs752444854 Chr.11: 106094535-106094535 on Build GRCh38
rs373397044 Chr.11: 106094665-106094665 on Build GRCh38
rs143179580 Chr.11: 106094646-106094646 on Build GRCh38
rs564320606 Chr.11: 106094644-106094644 on Build GRCh38
rs182606906 Chr.11: 106094500-106094500 on Build GRCh38
rs772230896 Chr.11: 106094596-106094596 on Build GRCh38

View Full Product Details