Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 27208084-27208433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACBD5
Entrez Gene ID
91452
Gene Name
acyl-CoA binding domain containing 5
Gene Aliases
-
UniGene
Hs.530597
Gene Chromosome Location
Chr.10: 27182624-27248674 on Build GRCh38
Transcripts
NM_001042473.3 NM_001271512.3 NM_001301251.1 NM_001301252.1 NM_001301253.1 NM_001301254.1 NM_145698.4 XM_006717528.3 XM_006717530.3 XM_006717531.2 XM_006717535.2 XM_017016884.1 XM_017016885.1 XM_017016886.1 XM_017016887.1 XM_017016888.1 XM_017016889.1 XM_017016890.1 XM_017016891.1 XM_017016892.1 XM_017016893.1 XM_017016894.1 XM_017016895.1 XM_017016896.1 XM_017016897.1 XM_017016898.1 XM_017016899.1 XM_017016900.1 XM_017016901.1 XM_017016902.1 XM_017016903.1 XM_017016904.1 XM_017016905.1 XM_017016906.1 XM_017016907.1 XM_017016908.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3358717 c.1204A>T p.N402Y Chr.10: 27208341-27208341 on Build GRCh38
4876913 c.1348G>T p.A450S Chr.10: 27208329-27208329 on Build GRCh38
7212712 c.1222G>A p.V408M Chr.10: 27208323-27208323 on Build GRCh38
6591882 c.1295C>A p.S432Y Chr.10: 27208355-27208355 on Build GRCh38
4421370 c.1304G>A p.S435N Chr.10: 27208346-27208346 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1046160854 Chr.10: 27208361-27208361 on Build GRCh38
rs748895600 Chr.10: 27208212-27208212 on Build GRCh38
rs189115180 Chr.10: 27208324-27208324 on Build GRCh38
rs779982287 Chr.10: 27208377-27208377 on Build GRCh38
rs768000744 Chr.10: 27208352-27208352 on Build GRCh38
rs746920760 Chr.10: 27208294-27208294 on Build GRCh38
rs776573195 Chr.10: 27208313-27208313 on Build GRCh38
rs778319933 Chr.10: 27208366-27208366 on Build GRCh38
rs755472165 Chr.10: 27208349-27208349 on Build GRCh38
rs768344753 Chr.10: 27208381-27208381 on Build GRCh38
rs561228953 Chr.10: 27208242-27208242 on Build GRCh38
rs560917434 Chr.10: 27208203-27208203 on Build GRCh38
rs771953638 Chr.10: 27208247-27208247 on Build GRCh38
rs7919523 Chr.10: 27208145-27208145 on Build GRCh38
rs1054523535 Chr.10: 27208387-27208387 on Build GRCh38
rs781128539 Chr.10: 27208382-27208382 on Build GRCh38
rs186201246 Chr.10: 27208118-27208118 on Build GRCh38
rs778569688 Chr.10: 27208229-27208229 on Build GRCh38
rs761888781 Chr.10: 27208315-27208315 on Build GRCh38
rs59672993 Chr.10: 27208143-27208143 on Build GRCh38
rs765248478 Chr.10: 27208319-27208319 on Build GRCh38
rs918553903 Chr.10: 27208237-27208237 on Build GRCh38
rs1027099238 Chr.10: 27208336-27208336 on Build GRCh38
rs181926234 Chr.10: 27208330-27208330 on Build GRCh38
rs756724312 Chr.10: 27208225-27208225 on Build GRCh38
rs544905257 Chr.10: 27208180-27208180 on Build GRCh38
rs553480972 Chr.10: 27208148-27208148 on Build GRCh38
rs140046853 Chr.10: 27208156-27208156 on Build GRCh38
rs77127632 Chr.10: 27208261-27208261 on Build GRCh38
rs769882516 Chr.10: 27208399-27208399 on Build GRCh38
rs148555426 Chr.10: 27208379-27208379 on Build GRCh38
rs774559584 Chr.10: 27208412-27208412 on Build GRCh38
rs745535917 Chr.10: 27208245-27208245 on Build GRCh38
rs773326165 Chr.10: 27208401-27208401 on Build GRCh38
rs758175108 Chr.10: 27208376-27208376 on Build GRCh38
rs7918793 Chr.10: 27208262-27208262 on Build GRCh38
rs369898700 Chr.10: 27208407-27208407 on Build GRCh38
rs751974758 Chr.10: 27208348-27208348 on Build GRCh38
rs777129981 Chr.10: 27208210-27208210 on Build GRCh38
rs753155998 Chr.10: 27208360-27208360 on Build GRCh38
rs1056013477 Chr.10: 27208323-27208323 on Build GRCh38
rs543720340 Chr.10: 27208406-27208406 on Build GRCh38
rs768701702 Chr.10: 27208300-27208300 on Build GRCh38
rs749980805 Chr.10: 27208359-27208360 on Build GRCh38
rs144860164 Chr.10: 27208365-27208365 on Build GRCh38
rs748060751 Chr.10: 27208392-27208392 on Build GRCh38
rs769561869 Chr.10: 27208112-27208112 on Build GRCh38
rs951430301 Chr.10: 27208259-27208259 on Build GRCh38
rs749212781 Chr.10: 27208329-27208329 on Build GRCh38
rs377281055 Chr.10: 27208371-27208371 on Build GRCh38
rs899033039 Chr.10: 27208326-27208326 on Build GRCh38

View Full Product Details