Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 121010909-121011212 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf43
Entrez Gene ID
64897
Gene Name
chromosome 12 open reading frame 43
Gene Aliases
-
UniGene
Hs.528671
Gene Chromosome Location
Chr.12: 121002422-121016502 on Build GRCh38
Transcripts
NM_001286191.1 NM_001286192.1 NM_001286195.1 NM_001286196.1 NM_001286197.1 NM_001286198.1 NM_022895.2 NR_104409.1 XM_017019829.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
340950 c.198G>A p.V66V Chr.12: 121010917-121010917 on Build GRCh38
5722062 c.173C>T p.S58F Chr.12: 121011119-121011119 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs148841072 Chr.12: 121011048-121011048 on Build GRCh38
rs756453529 Chr.12: 121011139-121011139 on Build GRCh38
rs769908111 Chr.12: 121011141-121011146 on Build GRCh38
rs781730210 Chr.12: 121011037-121011037 on Build GRCh38
rs961065462 Chr.12: 121011105-121011105 on Build GRCh38
rs751449265 Chr.12: 121010959-121010959 on Build GRCh38
rs143450063 Chr.12: 121011112-121011112 on Build GRCh38
rs915121825 Chr.12: 121010993-121010993 on Build GRCh38
rs11831650 Chr.12: 121011153-121011153 on Build GRCh38
rs763673604 Chr.12: 121011086-121011086 on Build GRCh38
rs770046713 Chr.12: 121010940-121010940 on Build GRCh38
rs756740298 Chr.12: 121011025-121011025 on Build GRCh38
rs199993208 Chr.12: 121011055-121011055 on Build GRCh38
rs934760541 Chr.12: 121011053-121011053 on Build GRCh38
rs753298773 Chr.12: 121011000-121011000 on Build GRCh38
rs764892159 Chr.12: 121011098-121011098 on Build GRCh38
rs1049737961 Chr.12: 121011089-121011089 on Build GRCh38
rs771128107 Chr.12: 121011171-121011171 on Build GRCh38
rs749565500 Chr.12: 121011165-121011165 on Build GRCh38
rs773427526 Chr.12: 121011049-121011049 on Build GRCh38
rs543590828 Chr.12: 121011046-121011046 on Build GRCh38
rs1002714214 Chr.12: 121011082-121011082 on Build GRCh38
rs753485465 Chr.12: 121011090-121011090 on Build GRCh38
rs759358022 Chr.12: 121010961-121010961 on Build GRCh38
rs749943738 Chr.12: 121011104-121011104 on Build GRCh38
rs777888699 Chr.12: 121011144-121011144 on Build GRCh38
rs936666072 Chr.12: 121010990-121010990 on Build GRCh38
rs764172699 Chr.12: 121010980-121010980 on Build GRCh38
rs773234722 Chr.12: 121010946-121010946 on Build GRCh38
rs755786668 Chr.12: 121010995-121010995 on Build GRCh38
rs959153876 Chr.12: 121011174-121011174 on Build GRCh38
rs770958518 Chr.12: 121011051-121011051 on Build GRCh38
rs536936192 Chr.12: 121011087-121011087 on Build GRCh38
rs761995376 Chr.12: 121010935-121010935 on Build GRCh38
rs747554465 Chr.12: 121011032-121011032 on Build GRCh38
rs756935269 Chr.12: 121011094-121011094 on Build GRCh38
rs778748854 Chr.12: 121011187-121011187 on Build GRCh38
rs769267169 Chr.12: 121011033-121011033 on Build GRCh38
rs780733128 Chr.12: 121011026-121011026 on Build GRCh38
rs567366188 Chr.12: 121010947-121010947 on Build GRCh38
rs528006588 Chr.12: 121011113-121011113 on Build GRCh38
rs777828557 Chr.12: 121011153-121011153 on Build GRCh38
rs944247285 Chr.12: 121010950-121010950 on Build GRCh38
rs777415707 Chr.12: 121010996-121010996 on Build GRCh38
rs766318791 Chr.12: 121010956-121010956 on Build GRCh38
rs76625631 Chr.12: 121011123-121011123 on Build GRCh38
rs201908756 Chr.12: 121011063-121011063 on Build GRCh38
rs376970818 Chr.12: 121010962-121010962 on Build GRCh38
rs760459871 Chr.12: 121011073-121011073 on Build GRCh38
rs112728225 Chr.12: 121011115-121011115 on Build GRCh38
rs752438120 Chr.12: 121011071-121011071 on Build GRCh38

View Full Product Details