Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10089559-10090108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf66
Entrez Gene ID
55337
Gene Name
chromosome 19 open reading frame 66
Gene Aliases
RyDEN
UniGene
Hs.655613
Gene Chromosome Location
Chr.19: 10086119-10093252 on Build GRCh38
Transcripts
NM_001308277.1 NM_018381.3 XM_011528121.1 XM_011528122.1 XM_017026934.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5706831 c.109A>G p.T37A Chr.19: 10089925-10089925 on Build GRCh38
5599166 c.341C>T p.S114F Chr.19: 10090004-10090004 on Build GRCh38
3528075 c.218C>T p.P73L Chr.19: 10090034-10090034 on Build GRCh38
5625208 c.290G>A p.R97H Chr.19: 10089953-10089953 on Build GRCh38
3528073 c.185C>T p.S62F Chr.19: 10090001-10090001 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377455252 Chr.19: 10089969-10089969 on Build GRCh38
rs930932259 Chr.19: 10089773-10089773 on Build GRCh38
rs369043885 Chr.19: 10090001-10090001 on Build GRCh38
rs772391880 Chr.19: 10089641-10089641 on Build GRCh38
rs745374621 Chr.19: 10089967-10089967 on Build GRCh38
rs1009795139 Chr.19: 10089856-10089856 on Build GRCh38
rs778737923 Chr.19: 10090056-10090056 on Build GRCh38
rs748924162 Chr.19: 10089739-10089739 on Build GRCh38
rs748989085 Chr.19: 10089959-10089959 on Build GRCh38
rs763222546 Chr.19: 10090002-10090002 on Build GRCh38
rs766605434 Chr.19: 10090009-10090009 on Build GRCh38
rs151207105 Chr.19: 10089637-10089637 on Build GRCh38
rs115304263 Chr.19: 10089830-10089830 on Build GRCh38
rs781665835 Chr.19: 10089718-10089718 on Build GRCh38
rs765381361 Chr.19: 10089680-10089680 on Build GRCh38
rs925037475 Chr.19: 10089713-10089713 on Build GRCh38
rs773757785 Chr.19: 10089861-10089862 on Build GRCh38
rs2232066 Chr.19: 10090085-10090085 on Build GRCh38
rs766509247 Chr.19: 10089912-10089912 on Build GRCh38
rs372496056 Chr.19: 10089714-10089714 on Build GRCh38
rs370815479 Chr.19: 10090029-10090029 on Build GRCh38
rs762540494 Chr.19: 10089978-10089979 on Build GRCh38
rs760635925 Chr.19: 10089981-10089981 on Build GRCh38
rs764433259 Chr.19: 10090030-10090030 on Build GRCh38
rs1009414241 Chr.19: 10089618-10089618 on Build GRCh38
rs758492353 Chr.19: 10089684-10089684 on Build GRCh38
rs775429263 Chr.19: 10089870-10089870 on Build GRCh38
rs752524600 Chr.19: 10089689-10089689 on Build GRCh38
rs372962161 Chr.19: 10090027-10090027 on Build GRCh38
rs369501193 Chr.19: 10089868-10089868 on Build GRCh38
rs755878880 Chr.19: 10089666-10089666 on Build GRCh38
rs1031970306 Chr.19: 10089946-10089946 on Build GRCh38
rs867282910 Chr.19: 10089811-10089811 on Build GRCh38
rs532630915 Chr.19: 10089790-10089790 on Build GRCh38
rs35325907 Chr.19: 10089733-10089734 on Build GRCh38
rs531330801 Chr.19: 10090044-10090044 on Build GRCh38
rs757891893 Chr.19: 10090071-10090071 on Build GRCh38
rs778179423 Chr.19: 10089952-10089952 on Build GRCh38
rs763756554 Chr.19: 10089599-10089599 on Build GRCh38
rs200888003 Chr.19: 10089661-10089661 on Build GRCh38
rs527346717 Chr.19: 10089841-10089841 on Build GRCh38
rs956670027 Chr.19: 10089964-10089964 on Build GRCh38
rs748477661 Chr.19: 10089719-10089719 on Build GRCh38
rs768371482 Chr.19: 10089984-10089984 on Build GRCh38
rs759646145 Chr.19: 10090025-10090025 on Build GRCh38
rs767057630 Chr.19: 10089880-10089880 on Build GRCh38
rs754228000 Chr.19: 10089882-10089914 on Build GRCh38
rs376814995 Chr.19: 10089778-10089779 on Build GRCh38
rs770704435 Chr.19: 10089960-10089960 on Build GRCh38
rs976207190 Chr.19: 10089999-10089999 on Build GRCh38
rs750658478 Chr.19: 10089893-10089893 on Build GRCh38
rs751717105 Chr.19: 10090015-10090015 on Build GRCh38
rs763127299 Chr.19: 10089906-10089906 on Build GRCh38
rs201891170 Chr.19: 10089914-10089914 on Build GRCh38
rs944870123 Chr.19: 10089736-10089736 on Build GRCh38
rs773968451 Chr.19: 10089864-10089864 on Build GRCh38
rs781558028 Chr.19: 10089924-10089924 on Build GRCh38
rs920844719 Chr.19: 10089740-10089740 on Build GRCh38
rs376516948 Chr.19: 10090028-10090028 on Build GRCh38
rs754267270 Chr.19: 10090039-10090039 on Build GRCh38
rs201429360 Chr.19: 10089941-10089941 on Build GRCh38
rs2232064 Chr.19: 10089726-10089726 on Build GRCh38
rs754997113 Chr.19: 10089918-10089918 on Build GRCh38
rs746981538 Chr.19: 10090086-10090086 on Build GRCh38
rs554005233 Chr.19: 10089600-10089600 on Build GRCh38
rs774454643 Chr.19: 10089607-10089607 on Build GRCh38
rs944898044 Chr.19: 10089985-10089985 on Build GRCh38
rs745467024 Chr.19: 10090068-10090068 on Build GRCh38
rs760417964 Chr.19: 10089877-10089877 on Build GRCh38
rs775309121 Chr.19: 10089978-10089978 on Build GRCh38
rs1045269393 Chr.19: 10090065-10090065 on Build GRCh38
rs2232065 Chr.19: 10089781-10089781 on Build GRCh38
rs373536863 Chr.19: 10089878-10089878 on Build GRCh38
rs542629527 Chr.19: 10089681-10089681 on Build GRCh38
rs887750452 Chr.19: 10090041-10090041 on Build GRCh38
rs932011454 Chr.19: 10089820-10089820 on Build GRCh38
rs139147326 Chr.19: 10089685-10089685 on Build GRCh38
rs781537905 Chr.19: 10089840-10089840 on Build GRCh38
rs767460462 Chr.19: 10089645-10089645 on Build GRCh38
rs896351969 Chr.19: 10089881-10089881 on Build GRCh38
rs567222699 Chr.19: 10089963-10089963 on Build GRCh38
rs887877538 Chr.19: 10089834-10089834 on Build GRCh38
rs759603800 Chr.19: 10089867-10089867 on Build GRCh38
rs541591126 Chr.19: 10089817-10089817 on Build GRCh38
rs762641966 Chr.19: 10089577-10089577 on Build GRCh38
rs753211447 Chr.19: 10089940-10089940 on Build GRCh38
rs926887844 Chr.19: 10090084-10090084 on Build GRCh38
rs770614665 Chr.19: 10089853-10089853 on Build GRCh38
rs190951509 Chr.19: 10089786-10089786 on Build GRCh38
rs761104658 Chr.19: 10089862-10089862 on Build GRCh38
rs777323863 Chr.19: 10089732-10089732 on Build GRCh38
rs1005134009 Chr.19: 10089580-10089580 on Build GRCh38
rs752097891 Chr.19: 10089704-10089704 on Build GRCh38
rs992042174 Chr.19: 10089653-10089653 on Build GRCh38
rs776478367 Chr.19: 10089986-10089986 on Build GRCh38

View Full Product Details