Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73783829-73784222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf78
Entrez Gene ID
388960
Gene Name
chromosome 2 open reading frame 78
Gene Aliases
COG5373, hCG1989538
UniGene
Hs.456833
Gene Chromosome Location
Chr.2: 73784189-73817148 on Build GRCh38
Transcripts
NM_001080474.1

SNP Detail

SNP ID Chromosome Location
rs997310981 Chr.2: 73783941-73783941 on Build GRCh38
rs562595304 Chr.2: 73784198-73784198 on Build GRCh38
rs34114095 Chr.2: 73784058-73784059 on Build GRCh38
rs2567597 Chr.2: 73783888-73783888 on Build GRCh38
rs557802116 Chr.2: 73783945-73783945 on Build GRCh38
rs930521287 Chr.2: 73783890-73783890 on Build GRCh38
rs116734533 Chr.2: 73783875-73783875 on Build GRCh38
rs982335688 Chr.2: 73784006-73784006 on Build GRCh38
rs535192295 Chr.2: 73783854-73783854 on Build GRCh38
rs552399457 Chr.2: 73783946-73783949 on Build GRCh38
rs190218005 Chr.2: 73783934-73783934 on Build GRCh38
rs1040332552 Chr.2: 73784161-73784161 on Build GRCh38
rs966901363 Chr.2: 73784001-73784001 on Build GRCh38
rs990960588 Chr.2: 73784053-73784053 on Build GRCh38
rs958429964 Chr.2: 73783946-73783946 on Build GRCh38
rs944566466 Chr.2: 73784151-73784151 on Build GRCh38
rs540949900 Chr.2: 73783968-73783968 on Build GRCh38
rs959080940 Chr.2: 73784038-73784038 on Build GRCh38
rs901157351 Chr.2: 73784200-73784200 on Build GRCh38
rs574215119 Chr.2: 73784193-73784193 on Build GRCh38
rs930468455 Chr.2: 73784084-73784084 on Build GRCh38
rs907716312 Chr.2: 73784137-73784137 on Build GRCh38
rs372271987 Chr.2: 73783919-73783919 on Build GRCh38
rs1021156891 Chr.2: 73783993-73783993 on Build GRCh38
rs886395317 Chr.2: 73783933-73783933 on Build GRCh38
rs920379879 Chr.2: 73784070-73784070 on Build GRCh38
rs181603664 Chr.2: 73783954-73783954 on Build GRCh38
rs927611705 Chr.2: 73784031-73784031 on Build GRCh38
rs544376139 Chr.2: 73784197-73784197 on Build GRCh38
rs920391706 Chr.2: 73783889-73783889 on Build GRCh38
rs184418630 Chr.2: 73783974-73783974 on Build GRCh38
rs901176544 Chr.2: 73783936-73783936 on Build GRCh38
rs1047628657 Chr.2: 73784125-73784125 on Build GRCh38

View Full Product Details