Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 72056467-72056902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf57
Entrez Gene ID
138240
Gene Name
chromosome 9 open reading frame 57
Gene Aliases
-
UniGene
Hs.371235
Gene Chromosome Location
Chr.9: 72051381-72060605 on Build GRCh38
Transcripts
NM_001128618.1 XM_011518225.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3721098 c.181A>G p.T61A Chr.9: 72056826-72056826 on Build GRCh38
3721100 c.184A>G p.K62E Chr.9: 72056823-72056823 on Build GRCh38
6851421 c.204A>G p.K68K Chr.9: 72056803-72056803 on Build GRCh38
5411213 c.223G>A p.D75N Chr.9: 72056784-72056784 on Build GRCh38
4971502 c.173C>T p.T58I Chr.9: 72056834-72056834 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs951711032 Chr.9: 72056567-72056567 on Build GRCh38
rs780644729 Chr.9: 72056832-72056832 on Build GRCh38
rs575697685 Chr.9: 72056641-72056641 on Build GRCh38
rs527622998 Chr.9: 72056750-72056750 on Build GRCh38
rs573300994 Chr.9: 72056602-72056602 on Build GRCh38
rs1055021046 Chr.9: 72056540-72056540 on Build GRCh38
rs530126679 Chr.9: 72056818-72056818 on Build GRCh38
rs555852469 Chr.9: 72056638-72056638 on Build GRCh38
rs545211242 Chr.9: 72056679-72056679 on Build GRCh38
rs1023499964 Chr.9: 72056810-72056810 on Build GRCh38
rs553772424 Chr.9: 72056595-72056595 on Build GRCh38
rs1031738886 Chr.9: 72056611-72056611 on Build GRCh38
rs771518272 Chr.9: 72056767-72056768 on Build GRCh38
rs972883983 Chr.9: 72056582-72056585 on Build GRCh38
rs140256268 Chr.9: 72056576-72056576 on Build GRCh38
rs762221670 Chr.9: 72056762-72056762 on Build GRCh38
rs756056608 Chr.9: 72056866-72056866 on Build GRCh38
rs966548661 Chr.9: 72056592-72056592 on Build GRCh38
rs927710552 Chr.9: 72056841-72056841 on Build GRCh38
rs999536150 Chr.9: 72056708-72056708 on Build GRCh38
rs200976346 Chr.9: 72056840-72056840 on Build GRCh38
rs1049967543 Chr.9: 72056596-72056596 on Build GRCh38
rs540964295 Chr.9: 72056802-72056802 on Build GRCh38
rs13295099 Chr.9: 72056803-72056803 on Build GRCh38
rs922737327 Chr.9: 72056593-72056593 on Build GRCh38
rs564910607 Chr.9: 72056709-72056709 on Build GRCh38
rs987886087 Chr.9: 72056666-72056666 on Build GRCh38
rs973714823 Chr.9: 72056780-72056780 on Build GRCh38
rs754510805 Chr.9: 72056826-72056826 on Build GRCh38
rs990672741 Chr.9: 72056514-72056514 on Build GRCh38
rs751158680 Chr.9: 72056823-72056823 on Build GRCh38
rs373557112 Chr.9: 72056844-72056844 on Build GRCh38
rs542171549 Chr.9: 72056614-72056614 on Build GRCh38
rs1010701041 Chr.9: 72056568-72056568 on Build GRCh38
rs961751915 Chr.9: 72056877-72056877 on Build GRCh38
rs566389702 Chr.9: 72056542-72056542 on Build GRCh38
rs890044851 Chr.9: 72056630-72056630 on Build GRCh38

View Full Product Details