Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 57506680-57507079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTCFL
Entrez Gene ID
140690
Gene Name
CCCTC-binding factor like
Gene Aliases
BORIS, CT27, CTCF-T, HMGB1L1, dJ579F20.2
UniGene
Hs.131543
Gene Chromosome Location
Chr.20: 57495965-57525652 on Build GRCh38
Transcripts
NM_001269040.1 NM_001269041.1 NM_001269042.1 NM_001269043.1 NM_001269044.1 NM_001269045.1 NM_001269046.1 NM_001269047.1 NM_001269048.1 NM_001269049.1 NM_001269050.1 NM_001269054.1 NM_001269055.1 NM_080618.3 NR_072975.1

SNP Detail

SNP ID Chromosome Location
rs909097278 Chr.20: 57506948-57506948 on Build GRCh38
rs535920079 Chr.20: 57506919-57506919 on Build GRCh38
rs6064554 Chr.20: 57507044-57507044 on Build GRCh38
rs934521365 Chr.20: 57506835-57506835 on Build GRCh38
rs915943311 Chr.20: 57506747-57506747 on Build GRCh38
rs1007290202 Chr.20: 57506894-57506894 on Build GRCh38
rs958860211 Chr.20: 57506731-57506731 on Build GRCh38
rs553073538 Chr.20: 57506974-57506974 on Build GRCh38
rs956690493 Chr.20: 57506876-57506876 on Build GRCh38
rs184107199 Chr.20: 57506754-57506754 on Build GRCh38
rs960098628 Chr.20: 57507012-57507012 on Build GRCh38
rs140704296 Chr.20: 57506874-57506874 on Build GRCh38
rs189185449 Chr.20: 57506824-57506824 on Build GRCh38
rs991608270 Chr.20: 57507013-57507013 on Build GRCh38
rs960006182 Chr.20: 57506711-57506711 on Build GRCh38
rs947286313 Chr.20: 57506758-57506758 on Build GRCh38
rs535056261 Chr.20: 57506722-57506722 on Build GRCh38
rs572747393 Chr.20: 57506982-57506982 on Build GRCh38
rs748737183 Chr.20: 57507011-57507012 on Build GRCh38
rs894654099 Chr.20: 57507040-57507040 on Build GRCh38
rs775187526 Chr.20: 57506815-57506815 on Build GRCh38
rs1013124061 Chr.20: 57506796-57506796 on Build GRCh38
rs556702804 Chr.20: 57506866-57506866 on Build GRCh38
rs372201250 Chr.20: 57506816-57506816 on Build GRCh38
rs967752630 Chr.20: 57506845-57506845 on Build GRCh38
rs1035884809 Chr.20: 57506989-57506989 on Build GRCh38
rs753944961 Chr.20: 57506977-57506977 on Build GRCh38
rs924402270 Chr.20: 57506827-57506827 on Build GRCh38
rs923140768 Chr.20: 57506914-57506914 on Build GRCh38
rs1057103573 Chr.20: 57507015-57507015 on Build GRCh38
rs996122822 Chr.20: 57506930-57506930 on Build GRCh38
rs1002951703 Chr.20: 57507007-57507007 on Build GRCh38
rs1036798575 Chr.20: 57506981-57506981 on Build GRCh38
rs1021797368 Chr.20: 57506806-57506806 on Build GRCh38
rs975822233 Chr.20: 57506901-57506901 on Build GRCh38
rs1050344353 Chr.20: 57506921-57506921 on Build GRCh38

View Full Product Details