Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 19324365-19324693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND4C
Entrez Gene ID
55667
Gene Name
DENN domain containing 4C
Gene Aliases
C9orf55, C9orf55B, RAB10GEF, bA513M16.3
UniGene
Hs.249591
Gene Chromosome Location
Chr.9: 19230765-19374268 on Build GRCh38
Transcripts
NM_017925.6 NR_073201.3 XM_006716804.3 XM_006716805.3 XM_006716807.3 XM_011517959.2 XM_017014917.1 XM_017014918.1 XM_017014919.1 XM_017014920.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5959555 c.1220C>A p.A407E Chr.9: 19324482-19324482 on Build GRCh38
1314705 c.1214G>A p.G405E Chr.9: 19324476-19324476 on Build GRCh38
3779913 c.1120C>T p.R374C Chr.9: 19324382-19324382 on Build GRCh38
4649324 c.1219G>A p.A407T Chr.9: 19324481-19324481 on Build GRCh38
6115187 c.1186T>C p.C396R Chr.9: 19324448-19324448 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745949702 Chr.9: 19324407-19324407 on Build GRCh38
rs781675431 Chr.9: 19324406-19324406 on Build GRCh38
rs760683218 Chr.9: 19324508-19324508 on Build GRCh38
rs146889923 Chr.9: 19324401-19324401 on Build GRCh38
rs369919342 Chr.9: 19324525-19324525 on Build GRCh38
rs539339101 Chr.9: 19324477-19324477 on Build GRCh38
rs376278355 Chr.9: 19324554-19324554 on Build GRCh38
rs748301824 Chr.9: 19324556-19324556 on Build GRCh38
rs1027094998 Chr.9: 19324657-19324657 on Build GRCh38
rs759288821 Chr.9: 19324528-19324528 on Build GRCh38
rs201341867 Chr.9: 19324526-19324526 on Build GRCh38
rs1045320833 Chr.9: 19324595-19324595 on Build GRCh38
rs768816127 Chr.9: 19324469-19324469 on Build GRCh38
rs765160034 Chr.9: 19324543-19324544 on Build GRCh38
rs776356130 Chr.9: 19324515-19324515 on Build GRCh38
rs892585543 Chr.9: 19324570-19324570 on Build GRCh38
rs200723777 Chr.9: 19324425-19324425 on Build GRCh38
rs536300374 Chr.9: 19324662-19324662 on Build GRCh38
rs751876009 Chr.9: 19324530-19324530 on Build GRCh38
rs756589357 Chr.9: 19324535-19324535 on Build GRCh38
rs762180638 Chr.9: 19324531-19324531 on Build GRCh38
rs905375659 Chr.9: 19324624-19324624 on Build GRCh38
rs1013849027 Chr.9: 19324571-19324571 on Build GRCh38
rs771967062 Chr.9: 19324559-19324559 on Build GRCh38
rs915541414 Chr.9: 19324502-19324502 on Build GRCh38
rs756245915 Chr.9: 19324414-19324414 on Build GRCh38
rs373856011 Chr.9: 19324547-19324547 on Build GRCh38
rs997571628 Chr.9: 19324649-19324649 on Build GRCh38
rs780128631 Chr.9: 19324545-19324545 on Build GRCh38
rs936864079 Chr.9: 19324549-19324549 on Build GRCh38
rs778604109 Chr.9: 19324473-19324473 on Build GRCh38
rs771913523 Chr.9: 19324483-19324483 on Build GRCh38
rs780075622 Chr.9: 19324433-19324433 on Build GRCh38
rs749560691 Chr.9: 19324461-19324461 on Build GRCh38
rs767761790 Chr.9: 19324532-19324532 on Build GRCh38
rs904095582 Chr.9: 19324546-19324546 on Build GRCh38
rs572606861 Chr.9: 19324575-19324575 on Build GRCh38
rs759478303 Chr.9: 19324538-19324538 on Build GRCh38
rs921770279 Chr.9: 19324472-19324472 on Build GRCh38
rs761686721 Chr.9: 19324400-19324400 on Build GRCh38
rs764605775 Chr.9: 19324391-19324391 on Build GRCh38
rs145319775 Chr.9: 19324495-19324495 on Build GRCh38
rs150040627 Chr.9: 19324402-19324402 on Build GRCh38

View Full Product Details