Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 86546866-86547195 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCA4
Entrez Gene ID
22802
Gene Name
chloride channel accessory 4
Gene Aliases
CaCC, CaCC2
UniGene
Hs.567422
Gene Chromosome Location
Chr.1: 86547076-86580758 on Build GRCh38
Transcripts
NM_012128.3 NR_024602.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6629796 c.50A>G p.H17R Chr.1: 86547169-86547169 on Build GRCh38
3419539 c.69C>A p.F23L Chr.1: 86547188-86547188 on Build GRCh38
6064618 c.66C>A p.S22S Chr.1: 86547185-86547185 on Build GRCh38
7361416 c.12C>A p.F4L Chr.1: 86547131-86547131 on Build GRCh38
4010354 c.46C>T p.L16L Chr.1: 86547165-86547165 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893528476 Chr.1: 86547152-86547152 on Build GRCh38
rs2231579 Chr.1: 86547103-86547103 on Build GRCh38
rs2231578 Chr.1: 86547045-86547045 on Build GRCh38
rs924484826 Chr.1: 86547043-86547043 on Build GRCh38
rs530092999 Chr.1: 86547161-86547162 on Build GRCh38
rs766512274 Chr.1: 86547163-86547163 on Build GRCh38
rs909927981 Chr.1: 86546998-86546998 on Build GRCh38
rs1009761072 Chr.1: 86547157-86547157 on Build GRCh38
rs746476631 Chr.1: 86547089-86547089 on Build GRCh38
rs201535476 Chr.1: 86547104-86547104 on Build GRCh38
rs543350050 Chr.1: 86546923-86546923 on Build GRCh38
rs751566278 Chr.1: 86547143-86547143 on Build GRCh38
rs759244513 Chr.1: 86547106-86547106 on Build GRCh38
rs769520992 Chr.1: 86547121-86547121 on Build GRCh38
rs755602775 Chr.1: 86547001-86547001 on Build GRCh38
rs1045531981 Chr.1: 86547018-86547018 on Build GRCh38
rs373534332 Chr.1: 86547068-86547068 on Build GRCh38
rs766948343 Chr.1: 86547109-86547109 on Build GRCh38
rs772296374 Chr.1: 86547149-86547149 on Build GRCh38
rs1007613315 Chr.1: 86547081-86547081 on Build GRCh38
rs950262266 Chr.1: 86547071-86547071 on Build GRCh38
rs752236551 Chr.1: 86547142-86547143 on Build GRCh38
rs201138221 Chr.1: 86547099-86547099 on Build GRCh38
rs1054578945 Chr.1: 86547046-86547046 on Build GRCh38
rs764003871 Chr.1: 86547136-86547136 on Build GRCh38
rs775267369 Chr.1: 86547124-86547124 on Build GRCh38
rs746222969 Chr.1: 86547110-86547110 on Build GRCh38
rs573742379 Chr.1: 86546961-86546961 on Build GRCh38
rs1028112351 Chr.1: 86546903-86546903 on Build GRCh38
rs55738496 Chr.1: 86546898-86546898 on Build GRCh38
rs149483374 Chr.1: 86546917-86546917 on Build GRCh38
rs1042039274 Chr.1: 86547162-86547162 on Build GRCh38
rs143875560 Chr.1: 86546922-86546922 on Build GRCh38
rs867938901 Chr.1: 86547007-86547007 on Build GRCh38
rs755267978 Chr.1: 86547168-86547168 on Build GRCh38
rs563528702 Chr.1: 86546940-86546940 on Build GRCh38
rs753957732 Chr.1: 86547165-86547165 on Build GRCh38
rs376362705 Chr.1: 86547088-86547088 on Build GRCh38
rs762801573 Chr.1: 86547129-86547129 on Build GRCh38

View Full Product Details