Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 8475987-8476284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC6A
Entrez Gene ID
93978
Gene Name
C-type lectin domain family 6 member A
Gene Aliases
CLEC4N, CLECSF10
UniGene
Hs.591147
Gene Chromosome Location
Chr.12: 8455995-8478330 on Build GRCh38
Transcripts
NM_001007033.1 NM_001317999.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6073857 c.464C>A p.T155K Chr.12: 8476219-8476219 on Build GRCh38
6138362 c.452_453GG>TT p.W151F Chr.12: 8476207-8476208 on Build GRCh38
347835 c.434G>T p.G145V Chr.12: 8476189-8476189 on Build GRCh38
4491159 c.375C>T p.F125F Chr.12: 8476130-8476130 on Build GRCh38
3813144 c.422C>T p.S141L Chr.12: 8476177-8476177 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770476646 Chr.12: 8476174-8476174 on Build GRCh38
rs767096285 Chr.12: 8476189-8476189 on Build GRCh38
rs765676874 Chr.12: 8476212-8476212 on Build GRCh38
rs1028970499 Chr.12: 8476041-8476041 on Build GRCh38
rs958916546 Chr.12: 8476191-8476191 on Build GRCh38
rs991687165 Chr.12: 8476210-8476210 on Build GRCh38
rs764772097 Chr.12: 8476072-8476072 on Build GRCh38
rs762732307 Chr.12: 8476173-8476173 on Build GRCh38
rs376244774 Chr.12: 8476087-8476087 on Build GRCh38
rs373627140 Chr.12: 8476182-8476182 on Build GRCh38
rs150726898 Chr.12: 8476217-8476217 on Build GRCh38
rs933215461 Chr.12: 8476011-8476011 on Build GRCh38
rs757671608 Chr.12: 8476103-8476103 on Build GRCh38
rs1040876504 Chr.12: 8476258-8476258 on Build GRCh38
rs770961292 Chr.12: 8476106-8476106 on Build GRCh38
rs771937363 Chr.12: 8476046-8476046 on Build GRCh38
rs1035714792 Chr.12: 8476169-8476169 on Build GRCh38
rs762488090 Chr.12: 8476076-8476076 on Build GRCh38
rs888758375 Chr.12: 8476069-8476069 on Build GRCh38
rs780272346 Chr.12: 8476224-8476224 on Build GRCh38
rs928906930 Chr.12: 8476172-8476172 on Build GRCh38
rs202005542 Chr.12: 8476241-8476241 on Build GRCh38
rs1020778246 Chr.12: 8476071-8476071 on Build GRCh38
rs763900288 Chr.12: 8476059-8476059 on Build GRCh38
rs781135039 Chr.12: 8476148-8476148 on Build GRCh38
rs139024188 Chr.12: 8476219-8476219 on Build GRCh38
rs779100677 Chr.12: 8476109-8476109 on Build GRCh38
rs746152441 Chr.12: 8476111-8476111 on Build GRCh38
rs371329334 Chr.12: 8476245-8476245 on Build GRCh38
rs369189992 Chr.12: 8476255-8476255 on Build GRCh38
rs756742845 Chr.12: 8476090-8476090 on Build GRCh38
rs564502509 Chr.12: 8476102-8476102 on Build GRCh38
rs769676852 Chr.12: 8476155-8476155 on Build GRCh38
rs113506277 Chr.12: 8476122-8476122 on Build GRCh38
rs751737591 Chr.12: 8476226-8476226 on Build GRCh38
rs200474786 Chr.12: 8476129-8476129 on Build GRCh38
rs775265136 Chr.12: 8476234-8476234 on Build GRCh38
rs762205996 Chr.12: 8476202-8476202 on Build GRCh38
rs754420545 Chr.12: 8476192-8476192 on Build GRCh38
rs772850683 Chr.12: 8476171-8476171 on Build GRCh38
rs368271517 Chr.12: 8476149-8476149 on Build GRCh38
rs749189565 Chr.12: 8476084-8476084 on Build GRCh38
rs755068526 Chr.12: 8476235-8476235 on Build GRCh38
rs768668679 Chr.12: 8476137-8476137 on Build GRCh38
rs908257644 Chr.12: 8476237-8476237 on Build GRCh38
rs764710113 Chr.12: 8476099-8476099 on Build GRCh38
rs759122331 Chr.12: 8476188-8476188 on Build GRCh38
rs774508722 Chr.12: 8476105-8476106 on Build GRCh38
rs758521576 Chr.12: 8476113-8476113 on Build GRCh38
rs765953064 Chr.12: 8476078-8476078 on Build GRCh38

View Full Product Details