Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90602062-90602391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTC3
Entrez Gene ID
64784
Gene Name
CREB regulated transcription coactivator 3
Gene Aliases
TORC-3, TORC3
UniGene
Hs.567572
Gene Chromosome Location
Chr.15: 90529886-90645345 on Build GRCh38
Transcripts
NM_001042574.2 NM_022769.4 XM_005254968.2 XM_011521906.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5696947 c.352-2A>G p.? Chr.15: 90602322-90602322 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760765679 Chr.15: 90602300-90602300 on Build GRCh38
rs533171234 Chr.15: 90602112-90602112 on Build GRCh38
rs569954961 Chr.15: 90602158-90602158 on Build GRCh38
rs961493609 Chr.15: 90602339-90602339 on Build GRCh38
rs764242720 Chr.15: 90602353-90602354 on Build GRCh38
rs370795243 Chr.15: 90602277-90602277 on Build GRCh38
rs772951699 Chr.15: 90602333-90602333 on Build GRCh38
rs571865686 Chr.15: 90602185-90602185 on Build GRCh38
rs551553155 Chr.15: 90602140-90602140 on Build GRCh38
rs1003185836 Chr.15: 90602294-90602294 on Build GRCh38
rs375325004 Chr.15: 90602349-90602349 on Build GRCh38
rs867140589 Chr.15: 90602095-90602095 on Build GRCh38
rs1022557774 Chr.15: 90602254-90602254 on Build GRCh38
rs901988654 Chr.15: 90602093-90602093 on Build GRCh38
rs911141549 Chr.15: 90602183-90602183 on Build GRCh38
rs972773477 Chr.15: 90602092-90602092 on Build GRCh38
rs774272980 Chr.15: 90602320-90602320 on Build GRCh38
rs1011291173 Chr.15: 90602212-90602212 on Build GRCh38
rs147681624 Chr.15: 90602327-90602327 on Build GRCh38
rs924925068 Chr.15: 90602249-90602249 on Build GRCh38
rs867725843 Chr.15: 90602242-90602242 on Build GRCh38
rs181380710 Chr.15: 90602354-90602354 on Build GRCh38
rs934881463 Chr.15: 90602138-90602138 on Build GRCh38
rs892850471 Chr.15: 90602165-90602165 on Build GRCh38
rs530681110 Chr.15: 90602219-90602219 on Build GRCh38
rs765883322 Chr.15: 90602355-90602355 on Build GRCh38
rs1043453815 Chr.15: 90602247-90602247 on Build GRCh38
rs879045574 Chr.15: 90602357-90602357 on Build GRCh38
rs775397089 Chr.15: 90602284-90602284 on Build GRCh38
rs747892988 Chr.15: 90602324-90602324 on Build GRCh38
rs549245812 Chr.15: 90602317-90602317 on Build GRCh38
rs8027641 Chr.15: 90602086-90602086 on Build GRCh38
rs1040645893 Chr.15: 90602087-90602087 on Build GRCh38
rs754591824 Chr.15: 90602184-90602184 on Build GRCh38

View Full Product Details