Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 29246759-29247268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYBA1
Entrez Gene ID
1411
Gene Name
crystallin beta A1
Gene Aliases
CRYB1, CTRCT10
UniGene
Hs.46275
Gene Chromosome Location
Chr.17: 29246857-29254494 on Build GRCh38
Transcripts
NM_005208.4 XM_017024198.1

SNP Detail

SNP ID Chromosome Location
rs752753329 Chr.17: 29246872-29246872 on Build GRCh38
rs759775425 Chr.17: 29246895-29246898 on Build GRCh38
rs79470827 Chr.17: 29246822-29246822 on Build GRCh38
rs949228821 Chr.17: 29246959-29246959 on Build GRCh38
rs527481790 Chr.17: 29247203-29247203 on Build GRCh38
rs1046688291 Chr.17: 29246964-29246964 on Build GRCh38
rs988030980 Chr.17: 29246947-29246947 on Build GRCh38
rs747366748 Chr.17: 29246898-29246898 on Build GRCh38
rs766505819 Chr.17: 29246856-29246856 on Build GRCh38
rs747708943 Chr.17: 29246924-29246924 on Build GRCh38
rs765316399 Chr.17: 29246868-29246868 on Build GRCh38
rs757547144 Chr.17: 29246901-29246901 on Build GRCh38
rs1015064541 Chr.17: 29246810-29246810 on Build GRCh38
rs1006479749 Chr.17: 29246800-29246800 on Build GRCh38
rs372586390 Chr.17: 29246916-29246916 on Build GRCh38
rs959179475 Chr.17: 29247113-29247133 on Build GRCh38
rs773352866 Chr.17: 29246938-29246938 on Build GRCh38
rs913713392 Chr.17: 29246956-29246956 on Build GRCh38
rs936553756 Chr.17: 29246936-29246936 on Build GRCh38
rs369560721 Chr.17: 29246857-29246857 on Build GRCh38
rs1021731123 Chr.17: 29247157-29247157 on Build GRCh38
rs548920481 Chr.17: 29247132-29247132 on Build GRCh38
rs748494738 Chr.17: 29247150-29247150 on Build GRCh38
rs748920321 Chr.17: 29246913-29246913 on Build GRCh38
rs562104647 Chr.17: 29246854-29246854 on Build GRCh38
rs773755994 Chr.17: 29246813-29246813 on Build GRCh38
rs376450228 Chr.17: 29246850-29246850 on Build GRCh38
rs778307276 Chr.17: 29246888-29246888 on Build GRCh38
rs112540623 Chr.17: 29247107-29247107 on Build GRCh38
rs1019787758 Chr.17: 29247069-29247069 on Build GRCh38
rs548077316 Chr.17: 29247126-29247126 on Build GRCh38
rs760693890 Chr.17: 29246944-29246944 on Build GRCh38
rs758875312 Chr.17: 29246874-29246874 on Build GRCh38
rs977430228 Chr.17: 29247183-29247183 on Build GRCh38
rs560440665 Chr.17: 29247186-29247186 on Build GRCh38
rs141269311 Chr.17: 29246863-29246863 on Build GRCh38
rs917783629 Chr.17: 29246837-29246837 on Build GRCh38
rs781419366 Chr.17: 29246912-29246912 on Build GRCh38
rs1006450660 Chr.17: 29247221-29247221 on Build GRCh38
rs746422034 Chr.17: 29247071-29247071 on Build GRCh38
rs907274264 Chr.17: 29246978-29246978 on Build GRCh38
rs887340974 Chr.17: 29247028-29247028 on Build GRCh38
rs772546953 Chr.17: 29247131-29247131 on Build GRCh38
rs779878639 Chr.17: 29247232-29247232 on Build GRCh38
rs199651551 Chr.17: 29246821-29246821 on Build GRCh38
rs969095681 Chr.17: 29247177-29247177 on Build GRCh38
rs937459272 Chr.17: 29246981-29246981 on Build GRCh38
rs887784060 Chr.17: 29247219-29247219 on Build GRCh38
rs200767305 Chr.17: 29246833-29246833 on Build GRCh38
rs779528022 Chr.17: 29246825-29246825 on Build GRCh38
rs772123294 Chr.17: 29246928-29246928 on Build GRCh38
rs773879352 Chr.17: 29246922-29246922 on Build GRCh38

View Full Product Details