Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23995023-23995376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS4L2
Entrez Gene ID
317749
Gene Name
dehydrogenase/reductase 4 like 2
Gene Aliases
SDR25C3
UniGene
Hs.743442
Gene Chromosome Location
Chr.14: 23969874-24006408 on Build GRCh38
Transcripts
NM_001193635.1 NM_001193636.1 NM_001193637.1 NM_198083.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3677937 c.359A>C p.N120T Chr.14: 23995084-23995084 on Build GRCh38
3773701 c.376C>A p.L126I Chr.14: 23995101-23995101 on Build GRCh38
5847891 c.376_377delCT p.L126fs*>30 Chr.14: 23995101-23995102 on Build GRCh38
3677941 c.380T>G p.M127R Chr.14: 23995105-23995105 on Build GRCh38
6261871 c.399G>T p.V133V Chr.14: 23995124-23995124 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368101183 Chr.14: 23995155-23995155 on Build GRCh38
rs971082673 Chr.14: 23995215-23995215 on Build GRCh38
rs151008126 Chr.14: 23995127-23995127 on Build GRCh38
rs927045690 Chr.14: 23995225-23995225 on Build GRCh38
rs753453952 Chr.14: 23995162-23995162 on Build GRCh38
rs148508271 Chr.14: 23995048-23995048 on Build GRCh38
rs770199686 Chr.14: 23995176-23995176 on Build GRCh38
rs61999853 Chr.14: 23995101-23995101 on Build GRCh38
rs1051054891 Chr.14: 23995272-23995272 on Build GRCh38
rs557070353 Chr.14: 23995074-23995074 on Build GRCh38
rs370782611 Chr.14: 23995055-23995055 on Build GRCh38
rs1039786940 Chr.14: 23995150-23995150 on Build GRCh38
rs771854139 Chr.14: 23995184-23995184 on Build GRCh38
rs747408253 Chr.14: 23995142-23995142 on Build GRCh38
rs1013879066 Chr.14: 23995190-23995190 on Build GRCh38
rs761499962 Chr.14: 23995164-23995164 on Build GRCh38
rs61999854 Chr.14: 23995165-23995165 on Build GRCh38
rs766372896 Chr.14: 23995077-23995077 on Build GRCh38
rs748800221 Chr.14: 23995051-23995051 on Build GRCh38
rs749398149 Chr.14: 23995181-23995181 on Build GRCh38
rs11556284 Chr.14: 23995115-23995115 on Build GRCh38
rs768704749 Chr.14: 23995145-23995145 on Build GRCh38
rs72692120 Chr.14: 23995156-23995156 on Build GRCh38
rs772289901 Chr.14: 23995065-23995065 on Build GRCh38
rs775968672 Chr.14: 23995069-23995069 on Build GRCh38
rs528798344 Chr.14: 23995249-23995249 on Build GRCh38
rs375882991 Chr.14: 23995171-23995171 on Build GRCh38
rs182017660 Chr.14: 23995341-23995341 on Build GRCh38
rs774462921 Chr.14: 23995081-23995081 on Build GRCh38
rs189352232 Chr.14: 23995085-23995085 on Build GRCh38
rs759560964 Chr.14: 23995084-23995084 on Build GRCh38
rs72692119 Chr.14: 23995151-23995151 on Build GRCh38
rs143083153 Chr.14: 23995128-23995128 on Build GRCh38
rs372909905 Chr.14: 23995170-23995170 on Build GRCh38
rs113886442 Chr.14: 23995073-23995073 on Build GRCh38
rs72692121 Chr.14: 23995276-23995276 on Build GRCh38
rs942721930 Chr.14: 23995285-23995285 on Build GRCh38
rs777462197 Chr.14: 23995139-23995139 on Build GRCh38
rs748200084 Chr.14: 23995154-23995154 on Build GRCh38
rs75291130 Chr.14: 23995214-23995214 on Build GRCh38
rs111637315 Chr.14: 23995257-23995257 on Build GRCh38
rs527247728 Chr.14: 23995328-23995328 on Build GRCh38
rs769275551 Chr.14: 23995136-23995137 on Build GRCh38
rs141465413 Chr.14: 23995099-23995099 on Build GRCh38
rs536430784 Chr.14: 23995329-23995329 on Build GRCh38
rs777997006 Chr.14: 23995177-23995177 on Build GRCh38
rs544859809 Chr.14: 23995284-23995284 on Build GRCh38
rs748361420 Chr.14: 23995174-23995174 on Build GRCh38
rs551327737 Chr.14: 23995264-23995264 on Build GRCh38
rs541400844 Chr.14: 23995132-23995132 on Build GRCh38
rs550395967 Chr.14: 23995307-23995307 on Build GRCh38
rs1051537255 Chr.14: 23995125-23995125 on Build GRCh38
rs752812789 Chr.14: 23995309-23995309 on Build GRCh38
rs149046158 Chr.14: 23995116-23995116 on Build GRCh38
rs918470242 Chr.14: 23995252-23995252 on Build GRCh38
rs141194106 Chr.14: 23995172-23995172 on Build GRCh38
rs1053032316 Chr.14: 23995312-23995312 on Build GRCh38
rs979827176 Chr.14: 23995216-23995216 on Build GRCh38
rs929879506 Chr.14: 23995271-23995271 on Build GRCh38
rs929958383 Chr.14: 23995118-23995118 on Build GRCh38
rs903956332 Chr.14: 23995306-23995306 on Build GRCh38
rs762000184 Chr.14: 23995075-23995075 on Build GRCh38
rs878945355 Chr.14: 23995297-23995297 on Build GRCh38
rs147830049 Chr.14: 23995169-23995169 on Build GRCh38
rs146629945 Chr.14: 23995056-23995056 on Build GRCh38
rs542839651 Chr.14: 23995198-23995198 on Build GRCh38
rs778245836 Chr.14: 23995319-23995319 on Build GRCh38
rs962544792 Chr.14: 23995247-23995247 on Build GRCh38
rs75370830 Chr.14: 23995305-23995305 on Build GRCh38
rs746187817 Chr.14: 23995061-23995061 on Build GRCh38
rs374464242 Chr.14: 23995157-23995157 on Build GRCh38

View Full Product Details