Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 59598496-59599076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX40
Entrez Gene ID
79665
Gene Name
DEAH-box helicase 40
Gene Aliases
ARG147, DDX40, PAD
UniGene
Hs.29403
Gene Chromosome Location
Chr.17: 59565525-59608352 on Build GRCh38
Transcripts
NM_001166301.1 NM_024612.4 XM_006722083.2 XM_011525253.2 XM_011525254.2 XM_017025080.1 XM_017025081.1 XM_017025082.1 XM_017025083.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3773827 c.1653T>C p.N551N Chr.17: 59598807-59598807 on Build GRCh38
3773828 c.1509T>C p.N503N Chr.17: 59598807-59598807 on Build GRCh38
982066 c.1675A>G p.I559V Chr.17: 59598829-59598829 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1042564552 Chr.17: 59598522-59598522 on Build GRCh38
rs1032116329 Chr.17: 59598555-59598555 on Build GRCh38
rs932638334 Chr.17: 59598775-59598775 on Build GRCh38
rs761969667 Chr.17: 59598855-59598856 on Build GRCh38
rs376151196 Chr.17: 59598824-59598824 on Build GRCh38
rs754330227 Chr.17: 59598859-59598859 on Build GRCh38
rs764699166 Chr.17: 59598856-59598856 on Build GRCh38
rs767847178 Chr.17: 59598806-59598806 on Build GRCh38
rs780812094 Chr.17: 59598885-59598885 on Build GRCh38
rs769156264 Chr.17: 59598893-59598893 on Build GRCh38
rs553673246 Chr.17: 59598909-59598909 on Build GRCh38
rs920050306 Chr.17: 59598704-59598704 on Build GRCh38
rs1011948653 Chr.17: 59598610-59598610 on Build GRCh38
rs557753815 Chr.17: 59598984-59598984 on Build GRCh38
rs1024659756 Chr.17: 59598961-59598961 on Build GRCh38
rs757730524 Chr.17: 59598861-59598861 on Build GRCh38
rs1025276536 Chr.17: 59598641-59598641 on Build GRCh38
rs769953564 Chr.17: 59598979-59598979 on Build GRCh38
rs983665424 Chr.17: 59598652-59598652 on Build GRCh38
rs2697415 Chr.17: 59598896-59598896 on Build GRCh38
rs902234840 Chr.17: 59598523-59598523 on Build GRCh38
rs2697414 Chr.17: 59599050-59599050 on Build GRCh38
rs747709875 Chr.17: 59598892-59598892 on Build GRCh38
rs1000602625 Chr.17: 59598547-59598547 on Build GRCh38
rs750723028 Chr.17: 59598862-59598862 on Build GRCh38
rs572111633 Chr.17: 59598916-59598916 on Build GRCh38
rs770662749 Chr.17: 59598781-59598781 on Build GRCh38
rs774240093 Chr.17: 59598787-59598787 on Build GRCh38
rs894902938 Chr.17: 59598938-59598938 on Build GRCh38
rs773001666 Chr.17: 59598743-59598743 on Build GRCh38
rs762847650 Chr.17: 59598779-59598779 on Build GRCh38
rs535297672 Chr.17: 59598908-59598908 on Build GRCh38
rs1014724602 Chr.17: 59598653-59598653 on Build GRCh38
rs759841316 Chr.17: 59598789-59598789 on Build GRCh38
rs894808460 Chr.17: 59598583-59598583 on Build GRCh38
rs545541222 Chr.17: 59598937-59598937 on Build GRCh38
rs970672135 Chr.17: 59598645-59598645 on Build GRCh38
rs139117700 Chr.17: 59598807-59598807 on Build GRCh38
rs556011688 Chr.17: 59598638-59598638 on Build GRCh38
rs754697292 Chr.17: 59598864-59598864 on Build GRCh38
rs974110510 Chr.17: 59598701-59598701 on Build GRCh38
rs769831185 Chr.17: 59598727-59598727 on Build GRCh38
rs1012286340 Chr.17: 59598952-59598952 on Build GRCh38
rs747655326 Chr.17: 59598697-59598697 on Build GRCh38
rs963212239 Chr.17: 59598666-59598666 on Build GRCh38

View Full Product Details