Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 76492026-76492315 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTX2
Entrez Gene ID
113878
Gene Name
deltex E3 ubiquitin ligase 2
Gene Aliases
RNF58
UniGene
Hs.187058
Gene Chromosome Location
Chr.7: 76461655-76505995 on Build GRCh38
Transcripts
NM_001102594.1 NM_001102595.1 NM_001102596.1 NM_020892.2 XM_005250131.1 XM_005250132.1 XM_011515772.1 XM_011515773.2 XM_011515774.1 XM_011515775.1 XM_017011723.1 XM_017011724.1 XM_017011725.1 XM_017011726.1 XM_017011727.1 XM_017011728.1 XM_017011729.1 XM_017011730.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6110641 c.1007C>A p.A336E Chr.7: 76492251-76492251 on Build GRCh38
453404 c.948C>T p.S316S Chr.7: 76492192-76492192 on Build GRCh38
3882463 c.1001C>A p.A334E Chr.7: 76492245-76492245 on Build GRCh38
3640671 c.929C>T p.P310L Chr.7: 76492173-76492173 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs199908903 Chr.7: 76492184-76492184 on Build GRCh38
rs71552450 Chr.7: 76492265-76492265 on Build GRCh38
rs538258625 Chr.7: 76492115-76492115 on Build GRCh38
rs997281250 Chr.7: 76492235-76492235 on Build GRCh38
rs760696777 Chr.7: 76492241-76492241 on Build GRCh38
rs571593144 Chr.7: 76492180-76492180 on Build GRCh38
rs747856154 Chr.7: 76492163-76492163 on Build GRCh38
rs148279131 Chr.7: 76492192-76492192 on Build GRCh38
rs766189525 Chr.7: 76492253-76492253 on Build GRCh38
rs565766204 Chr.7: 76492295-76492296 on Build GRCh38
rs572150355 Chr.7: 76492269-76492269 on Build GRCh38
rs768764013 Chr.7: 76492245-76492245 on Build GRCh38
rs746371706 Chr.7: 76492217-76492217 on Build GRCh38
rs527741783 Chr.7: 76492071-76492071 on Build GRCh38
rs751532127 Chr.7: 76492122-76492122 on Build GRCh38
rs553892445 Chr.7: 76492199-76492199 on Build GRCh38
rs936876407 Chr.7: 76492074-76492074 on Build GRCh38
rs772180849 Chr.7: 76492228-76492228 on Build GRCh38
rs774775418 Chr.7: 76492194-76492194 on Build GRCh38
rs866871441 Chr.7: 76492096-76492096 on Build GRCh38
rs766243282 Chr.7: 76492121-76492121 on Build GRCh38
rs756017195 Chr.7: 76492045-76492045 on Build GRCh38
rs776853130 Chr.7: 76492248-76492248 on Build GRCh38
rs749615626 Chr.7: 76492186-76492186 on Build GRCh38
rs1638154 Chr.7: 76492052-76492052 on Build GRCh38
rs780820968 Chr.7: 76492154-76492154 on Build GRCh38
rs143114796 Chr.7: 76492151-76492151 on Build GRCh38
rs750216576 Chr.7: 76492118-76492118 on Build GRCh38
rs758145884 Chr.7: 76492119-76492119 on Build GRCh38
rs549210409 Chr.7: 76492082-76492082 on Build GRCh38
rs767509314 Chr.7: 76492276-76492276 on Build GRCh38
rs775526701 Chr.7: 76492240-76492240 on Build GRCh38
rs567622394 Chr.7: 76492102-76492102 on Build GRCh38
rs762712606 Chr.7: 76492249-76492249 on Build GRCh38
rs747246711 Chr.7: 76492286-76492287 on Build GRCh38
rs901562251 Chr.7: 76492204-76492204 on Build GRCh38

View Full Product Details