Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 14920816-14921054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERP27
Entrez Gene ID
121506
Gene Name
endoplasmic reticulum protein 27
Gene Aliases
C12orf46, PDIA8
UniGene
Hs.162143
Gene Chromosome Location
Chr.12: 14914027-14938549 on Build GRCh38
Transcripts
NM_001300784.1 NM_152321.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3416673 c.346C>G p.Q116E Chr.12: 14921036-14921036 on Build GRCh38
937580 c.342T>C p.N114N Chr.12: 14921040-14921040 on Build GRCh38
4040592 c.387C>T p.A129A Chr.12: 14920995-14920995 on Build GRCh38
2003134 c.389C>T p.T130I Chr.12: 14920993-14920993 on Build GRCh38
1163588 c.399C>T p.S133S Chr.12: 14920983-14920983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550297029 Chr.12: 14920889-14920889 on Build GRCh38
rs772143608 Chr.12: 14920965-14920965 on Build GRCh38
rs760521851 Chr.12: 14920970-14920970 on Build GRCh38
rs748957062 Chr.12: 14921026-14921026 on Build GRCh38
rs35042193 Chr.12: 14921018-14921018 on Build GRCh38
rs559202744 Chr.12: 14920942-14920942 on Build GRCh38
rs375689074 Chr.12: 14920989-14920989 on Build GRCh38
rs768209206 Chr.12: 14920952-14920952 on Build GRCh38
rs766741706 Chr.12: 14920894-14920894 on Build GRCh38
rs779660780 Chr.12: 14920939-14920939 on Build GRCh38
rs974380655 Chr.12: 14920986-14920986 on Build GRCh38
rs765302409 Chr.12: 14920992-14920992 on Build GRCh38
rs753230012 Chr.12: 14920916-14920916 on Build GRCh38
rs773791180 Chr.12: 14920957-14920957 on Build GRCh38
rs908246827 Chr.12: 14920961-14920961 on Build GRCh38
rs759652099 Chr.12: 14920917-14920917 on Build GRCh38
rs758861197 Chr.12: 14921012-14921012 on Build GRCh38
rs758842201 Chr.12: 14920919-14920919 on Build GRCh38
rs199978046 Chr.12: 14921019-14921019 on Build GRCh38
rs181363870 Chr.12: 14920885-14920885 on Build GRCh38
rs752722705 Chr.12: 14921002-14921002 on Build GRCh38
rs755051499 Chr.12: 14920899-14920899 on Build GRCh38
rs369504838 Chr.12: 14920998-14920998 on Build GRCh38
rs372760396 Chr.12: 14920981-14920981 on Build GRCh38
rs2445382 Chr.12: 14920897-14920897 on Build GRCh38
rs767578492 Chr.12: 14920946-14920947 on Build GRCh38
rs149706342 Chr.12: 14920982-14920982 on Build GRCh38
rs571098405 Chr.12: 14921011-14921011 on Build GRCh38
rs766313762 Chr.12: 14920973-14920973 on Build GRCh38
rs958587494 Chr.12: 14920867-14920867 on Build GRCh38
rs371311041 Chr.12: 14920909-14920909 on Build GRCh38
rs750260522 Chr.12: 14920888-14920888 on Build GRCh38
rs368533335 Chr.12: 14920926-14920926 on Build GRCh38
rs200481421 Chr.12: 14920955-14920955 on Build GRCh38
rs375526764 Chr.12: 14920923-14920923 on Build GRCh38
rs761756086 Chr.12: 14920964-14920964 on Build GRCh38
rs773105045 Chr.12: 14920968-14920968 on Build GRCh38
rs12580037 Chr.12: 14920922-14920922 on Build GRCh38
rs188846095 Chr.12: 14921025-14921025 on Build GRCh38
rs757588305 Chr.12: 14921024-14921024 on Build GRCh38

View Full Product Details