Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 45751265-45751615 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM21C
Entrez Gene ID
253725
Gene Name
family with sequence similarity 21 member C
Gene Aliases
FAM21A, VPEF
UniGene
Hs.365286
Gene Chromosome Location
Chr.10: 45727170-45792964 on Build GRCh38
Transcripts
NM_001169106.1 NM_001169107.1 NM_015262.2 XM_011539566.1 XM_011539567.1 XM_011539568.1 XM_011539569.1 XM_011539570.2 XM_011539571.1 XM_011539572.1 XM_017016014.1 XM_017016015.1 XM_017016016.1 XM_017016017.1 XM_017016018.1 XM_017016019.1 XM_017016020.1 XM_017016021.1 XM_017016022.1 XM_017016023.1 XM_017016025.1 XM_017016026.1

SNP Detail

SNP ID Chromosome Location
rs567512352 Chr.10: 45751518-45751518 on Build GRCh38
rs551723569 Chr.10: 45751404-45751404 on Build GRCh38
rs2257475 Chr.10: 45751337-45751337 on Build GRCh38
rs193206009 Chr.10: 45751341-45751341 on Build GRCh38
rs781980524 Chr.10: 45751574-45751574 on Build GRCh38
rs188766866 Chr.10: 45751322-45751322 on Build GRCh38
rs782219223 Chr.10: 45751512-45751512 on Build GRCh38
rs530867481 Chr.10: 45751447-45751447 on Build GRCh38
rs372618712 Chr.10: 45751553-45751553 on Build GRCh38
rs535258926 Chr.10: 45751549-45751549 on Build GRCh38
rs534000953 Chr.10: 45751351-45751351 on Build GRCh38
rs782680869 Chr.10: 45751456-45751456 on Build GRCh38
rs782259199 Chr.10: 45751505-45751505 on Build GRCh38
rs782376645 Chr.10: 45751508-45751508 on Build GRCh38
rs563352335 Chr.10: 45751431-45751431 on Build GRCh38
rs368686236 Chr.10: 45751291-45751291 on Build GRCh38
rs782028109 Chr.10: 45751537-45751537 on Build GRCh38
rs782551969 Chr.10: 45751453-45751453 on Build GRCh38
rs549114050 Chr.10: 45751509-45751509 on Build GRCh38

View Full Product Details