Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 45784524-45785002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM21C
Entrez Gene ID
253725
Gene Name
family with sequence similarity 21 member C
Gene Aliases
FAM21A, VPEF
UniGene
Hs.365286
Gene Chromosome Location
Chr.10: 45727170-45792964 on Build GRCh38
Transcripts
NM_001169106.1 NM_001169107.1 NM_015262.2 XM_011539566.1 XM_011539567.1 XM_011539568.1 XM_011539569.1 XM_011539570.2 XM_011539571.1 XM_011539573.2 XM_017016014.1 XM_017016015.1 XM_017016016.1 XM_017016017.1 XM_017016018.1 XM_017016019.1 XM_017016020.1 XM_017016021.1 XM_017016022.1 XM_017016023.1 XM_017016024.1 XM_017016025.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
918319 c.2481C>A p.G827G Chr.10: 45784567-45784567 on Build GRCh38
918320 c.2488C>T p.P830S Chr.10: 45784574-45784574 on Build GRCh38
397398 c.2620A>C p.S874R Chr.10: 45784833-45784833 on Build GRCh38
272665 c.2484C>T p.C828C Chr.10: 45784570-45784570 on Build GRCh38
918321 c.2617C>T p.P873S Chr.10: 45784830-45784830 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781877298 Chr.10: 45784589-45784589 on Build GRCh38
rs559892361 Chr.10: 45784733-45784733 on Build GRCh38
rs150463881 Chr.10: 45784963-45784963 on Build GRCh38
rs541566659 Chr.10: 45784732-45784732 on Build GRCh38
rs544156897 Chr.10: 45784570-45784570 on Build GRCh38
rs781802255 Chr.10: 45784614-45784614 on Build GRCh38
rs562350720 Chr.10: 45784650-45784650 on Build GRCh38
rs782736580 Chr.10: 45784613-45784613 on Build GRCh38
rs782452120 Chr.10: 45784899-45784899 on Build GRCh38
rs782398477 Chr.10: 45784929-45784929 on Build GRCh38
rs782082941 Chr.10: 45784566-45784566 on Build GRCh38
rs782681209 Chr.10: 45784637-45784637 on Build GRCh38
rs374325112 Chr.10: 45784949-45784949 on Build GRCh38
rs782383116 Chr.10: 45784806-45784806 on Build GRCh38
rs2597017 Chr.10: 45784568-45784568 on Build GRCh38
rs782059714 Chr.10: 45784834-45784834 on Build GRCh38
rs782750272 Chr.10: 45784648-45784649 on Build GRCh38
rs781808628 Chr.10: 45784891-45784891 on Build GRCh38
rs782239985 Chr.10: 45784944-45784944 on Build GRCh38
rs782285540 Chr.10: 45784923-45784923 on Build GRCh38
rs782626271 Chr.10: 45784803-45784803 on Build GRCh38
rs782611170 Chr.10: 45784880-45784880 on Build GRCh38
rs782684115 Chr.10: 45784838-45784838 on Build GRCh38
rs782625953 Chr.10: 45784680-45784680 on Build GRCh38
rs564363047 Chr.10: 45784783-45784783 on Build GRCh38
rs11499041 Chr.10: 45784738-45784738 on Build GRCh38
rs782431514 Chr.10: 45784625-45784625 on Build GRCh38
rs531509679 Chr.10: 45784829-45784829 on Build GRCh38
rs372605608 Chr.10: 45784922-45784922 on Build GRCh38
rs782147848 Chr.10: 45784849-45784849 on Build GRCh38
rs574192995 Chr.10: 45784673-45784673 on Build GRCh38
rs782445280 Chr.10: 45784604-45784604 on Build GRCh38
rs550948634 Chr.10: 45784863-45784863 on Build GRCh38
rs782227242 Chr.10: 45784688-45784688 on Build GRCh38
rs577483646 Chr.10: 45784555-45784555 on Build GRCh38
rs552099773 Chr.10: 45784755-45784755 on Build GRCh38
rs782778043 Chr.10: 45784854-45784854 on Build GRCh38
rs782172284 Chr.10: 45784586-45784586 on Build GRCh38
rs782428357 Chr.10: 45784779-45784779 on Build GRCh38
rs782140814 Chr.10: 45784804-45784804 on Build GRCh38
rs781923688 Chr.10: 45784571-45784571 on Build GRCh38
rs781968329 Chr.10: 45784557-45784557 on Build GRCh38
rs781829169 Chr.10: 45784661-45784662 on Build GRCh38
rs782799603 Chr.10: 45784587-45784587 on Build GRCh38
rs782340419 Chr.10: 45784720-45784720 on Build GRCh38
rs781899630 Chr.10: 45784845-45784845 on Build GRCh38
rs368543226 Chr.10: 45784598-45784598 on Build GRCh38
rs782640673 Chr.10: 45784937-45784937 on Build GRCh38

View Full Product Details