Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 61200461-61201011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHIT
Entrez Gene ID
2272
Gene Name
fragile histidine triad
Gene Aliases
AP3Aase, FRA3B
UniGene
Hs.655995
Gene Chromosome Location
Chr.3: 59747278-61251474 on Build GRCh38
Transcripts
NM_001166243.2 NM_001320899.1 NM_001320900.1 NM_002012.3 XM_017005880.1 XM_017005881.1 XM_017005882.1 XM_017005883.1 XM_017005884.1

SNP Detail

SNP ID Chromosome Location
rs148197698 Chr.3: 61200761-61200761 on Build GRCh38
rs1038498318 Chr.3: 61200499-61200499 on Build GRCh38
rs768926938 Chr.3: 61200921-61200921 on Build GRCh38
rs941440136 Chr.3: 61200709-61200709 on Build GRCh38
rs1030089013 Chr.3: 61200705-61200705 on Build GRCh38
rs758086241 Chr.3: 61200631-61200631 on Build GRCh38
rs554699623 Chr.3: 61200490-61200490 on Build GRCh38
rs556101253 Chr.3: 61200842-61200842 on Build GRCh38
rs774891279 Chr.3: 61200707-61200709 on Build GRCh38
rs532122462 Chr.3: 61200550-61200550 on Build GRCh38
rs993295075 Chr.3: 61200688-61200688 on Build GRCh38
rs746202195 Chr.3: 61200868-61200868 on Build GRCh38
rs957941027 Chr.3: 61200759-61200759 on Build GRCh38
rs114546131 Chr.3: 61200517-61200517 on Build GRCh38
rs942016626 Chr.3: 61200819-61200819 on Build GRCh38
rs989068738 Chr.3: 61200890-61200890 on Build GRCh38
rs115110347 Chr.3: 61200599-61200599 on Build GRCh38
rs77001996 Chr.3: 61200954-61200954 on Build GRCh38
rs1006157752 Chr.3: 61200624-61200624 on Build GRCh38
rs928738470 Chr.3: 61200798-61200798 on Build GRCh38
rs774289750 Chr.3: 61200947-61200947 on Build GRCh38
rs954461731 Chr.3: 61200708-61200708 on Build GRCh38
rs993348996 Chr.3: 61200857-61200857 on Build GRCh38
rs1033411970 Chr.3: 61200823-61200823 on Build GRCh38
rs540081433 Chr.3: 61200917-61200917 on Build GRCh38
rs1033505119 Chr.3: 61200601-61200601 on Build GRCh38
rs73097006 Chr.3: 61200521-61200521 on Build GRCh38
rs190683296 Chr.3: 61200605-61200605 on Build GRCh38
rs572976644 Chr.3: 61200889-61200889 on Build GRCh38
rs149431460 Chr.3: 61200512-61200512 on Build GRCh38
rs772724195 Chr.3: 61200539-61200539 on Build GRCh38
rs9846073 Chr.3: 61200876-61200876 on Build GRCh38
rs147372944 Chr.3: 61200956-61200956 on Build GRCh38
rs116620701 Chr.3: 61200638-61200638 on Build GRCh38
rs932759353 Chr.3: 61200949-61200949 on Build GRCh38
rs941471000 Chr.3: 61200496-61200496 on Build GRCh38
rs543451612 Chr.3: 61200941-61200941 on Build GRCh38
rs746600223 Chr.3: 61200741-61200741 on Build GRCh38
rs928841300 Chr.3: 61200948-61200948 on Build GRCh38
rs778674660 Chr.3: 61200527-61200527 on Build GRCh38
rs1016141575 Chr.3: 61200896-61200896 on Build GRCh38
rs762007277 Chr.3: 61200975-61200975 on Build GRCh38
rs537841801 Chr.3: 61200822-61200822 on Build GRCh38
rs967773002 Chr.3: 61200571-61200571 on Build GRCh38
rs891392051 Chr.3: 61200528-61200528 on Build GRCh38
rs371574715 Chr.3: 61200492-61200492 on Build GRCh38
rs377284007 Chr.3: 61200875-61200875 on Build GRCh38
rs370521464 Chr.3: 61200766-61200766 on Build GRCh38
rs181635131 Chr.3: 61200482-61200482 on Build GRCh38
rs986969404 Chr.3: 61200953-61200953 on Build GRCh38
rs114402059 Chr.3: 61200881-61200881 on Build GRCh38
rs1056262204 Chr.3: 61200814-61200814 on Build GRCh38
rs917812028 Chr.3: 61200651-61200651 on Build GRCh38
rs946223129 Chr.3: 61200768-61200768 on Build GRCh38
rs961905485 Chr.3: 61200655-61200655 on Build GRCh38
rs1055771780 Chr.3: 61200566-61200566 on Build GRCh38
rs959395203 Chr.3: 61200824-61200824 on Build GRCh38
rs141168938 Chr.3: 61200815-61200815 on Build GRCh38
rs796651300 Chr.3: 61200666-61200666 on Build GRCh38
rs1030141233 Chr.3: 61200869-61200869 on Build GRCh38
rs1020661730 Chr.3: 61200561-61200561 on Build GRCh38
rs1042073206 Chr.3: 61200807-61200807 on Build GRCh38
rs942043618 Chr.3: 61200958-61200958 on Build GRCh38
rs17064630 Chr.3: 61200821-61200821 on Build GRCh38
rs914782094 Chr.3: 61200483-61200483 on Build GRCh38
rs891365707 Chr.3: 61200805-61200805 on Build GRCh38
rs143857028 Chr.3: 61200736-61200736 on Build GRCh38
rs35198002 Chr.3: 61200806-61200806 on Build GRCh38
rs562439476 Chr.3: 61200957-61200957 on Build GRCh38
rs959196234 Chr.3: 61200617-61200617 on Build GRCh38
rs764681433 Chr.3: 61200665-61200665 on Build GRCh38
rs567021298 Chr.3: 61200776-61200776 on Build GRCh38

View Full Product Details