Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 48174453-48174902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMPD2
Entrez Gene ID
143162
Gene Name
FERM and PDZ domain containing 2
Gene Aliases
PDZD5C, PDZK4, PDZK5C
UniGene
Hs.664786
Gene Chromosome Location
Chr.10: 48156559-48274898 on Build GRCh38
Transcripts
NM_001018071.3 NM_001042512.2 NM_001318191.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3978551 c.3045G>A p.Q1015Q Chr.10: 48174900-48174900 on Build GRCh38
4144719 c.3062C>A p.T1021K Chr.10: 48174883-48174883 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs587734192 Chr.10: 48174747-48174747 on Build GRCh38
rs561680035 Chr.10: 48174787-48174787 on Build GRCh38
rs561333986 Chr.10: 48174762-48174762 on Build GRCh38
rs530414313 Chr.10: 48174799-48174799 on Build GRCh38
rs781786564 Chr.10: 48174824-48174824 on Build GRCh38
rs587613689 Chr.10: 48174755-48174755 on Build GRCh38
rs782472193 Chr.10: 48174730-48174730 on Build GRCh38
rs531155201 Chr.10: 48174545-48174545 on Build GRCh38
rs371077940 Chr.10: 48174738-48174738 on Build GRCh38
rs2564826 Chr.10: 48174516-48174516 on Build GRCh38
rs550192493 Chr.10: 48174841-48174841 on Build GRCh38
rs782246706 Chr.10: 48174847-48174847 on Build GRCh38
rs543295542 Chr.10: 48174630-48174630 on Build GRCh38
rs191864539 Chr.10: 48174507-48174507 on Build GRCh38
rs528639775 Chr.10: 48174767-48174767 on Build GRCh38
rs782451555 Chr.10: 48174832-48174832 on Build GRCh38
rs1898784 Chr.10: 48174883-48174883 on Build GRCh38
rs563651616 Chr.10: 48174529-48174529 on Build GRCh38
rs546312483 Chr.10: 48174772-48174772 on Build GRCh38
rs545452271 Chr.10: 48174520-48174520 on Build GRCh38
rs782629343 Chr.10: 48174814-48174814 on Build GRCh38

View Full Product Details