Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 154889223-154889473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GEMIN5
Entrez Gene ID
25929
Gene Name
gem nuclear organelle associated protein 5
Gene Aliases
GEMIN-5
UniGene
Hs.483921
Gene Chromosome Location
Chr.5: 154887416-154938216 on Build GRCh38
Transcripts
NM_001252156.1 NM_015465.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
23518 c.4351_4352delAG p.S1451fs*2 Chr.5: 154889328-154889329 on Build GRCh38
1208099 c.4348G>A p.E1450K Chr.5: 154889332-154889332 on Build GRCh38
6670451 c.4346C>A p.P1449H Chr.5: 154889334-154889334 on Build GRCh38
6027168 c.4289C>A p.S1430Y Chr.5: 154889391-154889391 on Build GRCh38
3245727 c.4322C>T p.A1441V Chr.5: 154889358-154889358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750658301 Chr.5: 154889396-154889396 on Build GRCh38
rs372810279 Chr.5: 154889446-154889446 on Build GRCh38
rs771760575 Chr.5: 154889439-154889439 on Build GRCh38
rs377532744 Chr.5: 154889280-154889280 on Build GRCh38
rs76192856 Chr.5: 154889392-154889392 on Build GRCh38
rs757932154 Chr.5: 154889363-154889363 on Build GRCh38
rs758445924 Chr.5: 154889305-154889305 on Build GRCh38
rs770757385 Chr.5: 154889353-154889353 on Build GRCh38
rs764656724 Chr.5: 154889279-154889279 on Build GRCh38
rs751474870 Chr.5: 154889437-154889437 on Build GRCh38
rs765602845 Chr.5: 154889394-154889394 on Build GRCh38
rs758665862 Chr.5: 154889397-154889397 on Build GRCh38
rs763708097 Chr.5: 154889425-154889425 on Build GRCh38
rs777756524 Chr.5: 154889407-154889407 on Build GRCh38
rs767989826 Chr.5: 154889423-154889424 on Build GRCh38
rs376067368 Chr.5: 154889328-154889328 on Build GRCh38
rs182269175 Chr.5: 154889401-154889401 on Build GRCh38
rs142314227 Chr.5: 154889321-154889321 on Build GRCh38
rs760617888 Chr.5: 154889371-154889371 on Build GRCh38
rs990945636 Chr.5: 154889289-154889289 on Build GRCh38
rs141457910 Chr.5: 154889365-154889365 on Build GRCh38
rs538483357 Chr.5: 154889307-154889307 on Build GRCh38
rs190950517 Chr.5: 154889301-154889301 on Build GRCh38
rs773917629 Chr.5: 154889358-154889358 on Build GRCh38
rs747037689 Chr.5: 154889326-154889326 on Build GRCh38
rs775172009 Chr.5: 154889364-154889364 on Build GRCh38
rs749220897 Chr.5: 154889409-154889409 on Build GRCh38
rs577227116 Chr.5: 154889427-154889427 on Build GRCh38
rs1053907643 Chr.5: 154889391-154889391 on Build GRCh38
rs544452250 Chr.5: 154889383-154889383 on Build GRCh38
rs753306593 Chr.5: 154889270-154889270 on Build GRCh38
rs753388051 Chr.5: 154889428-154889431 on Build GRCh38
rs112323495 Chr.5: 154889331-154889331 on Build GRCh38
rs779060051 Chr.5: 154889300-154889300 on Build GRCh38
rs370479756 Chr.5: 154889362-154889362 on Build GRCh38
rs757051838 Chr.5: 154889421-154889421 on Build GRCh38
rs816739 Chr.5: 154889342-154889342 on Build GRCh38
rs757583460 Chr.5: 154889285-154889285 on Build GRCh38
rs930260752 Chr.5: 154889359-154889359 on Build GRCh38
rs878857535 Chr.5: 154889268-154889268 on Build GRCh38
rs540609069 Chr.5: 154889403-154889403 on Build GRCh38
rs865946787 Chr.5: 154889335-154889335 on Build GRCh38
rs552624344 Chr.5: 154889271-154889271 on Build GRCh38
rs778792493 Chr.5: 154889431-154889431 on Build GRCh38

View Full Product Details