Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 99554374-99554513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFF3
Entrez Gene ID
3899
Gene Name
AF4/FMR2 family member 3
Gene Aliases
LAF4, MLLT2-like
UniGene
Hs.444414
Gene Chromosome Location
Chr.2: 99545861-100142583 on Build GRCh38
Transcripts
NM_001025108.1 NM_002285.2 XM_005263943.3 XM_005263945.3 XM_011511169.2 XM_011511170.2 XM_011511171.2 XM_011511172.2 XM_011511173.2 XM_011511174.2 XM_011511175.2 XM_011511176.2 XM_011511177.2 XM_011511178.2 XM_011511179.2 XM_017004085.1 XM_017004086.1 XM_017004087.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5991415 c.3546G>T p.M1182I Chr.2: 99554399-99554399 on Build GRCh38
5575612 c.3469T>C p.S1157P Chr.2: 99554476-99554476 on Build GRCh38
3396433 c.3497C>T p.A1166V Chr.2: 99554448-99554448 on Build GRCh38
715022 c.3533G>A p.R1178H Chr.2: 99554412-99554412 on Build GRCh38
6595895 c.3532C>T p.R1178C Chr.2: 99554413-99554413 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767846193 Chr.2: 99554406-99554406 on Build GRCh38
rs780887781 Chr.2: 99554456-99554456 on Build GRCh38
rs143764541 Chr.2: 99554482-99554482 on Build GRCh38
rs760851681 Chr.2: 99554492-99554492 on Build GRCh38
rs142364775 Chr.2: 99554410-99554410 on Build GRCh38
rs745505866 Chr.2: 99554458-99554458 on Build GRCh38
rs766758032 Chr.2: 99554412-99554412 on Build GRCh38
rs371101321 Chr.2: 99554471-99554471 on Build GRCh38
rs1015661536 Chr.2: 99554462-99554462 on Build GRCh38
rs773572168 Chr.2: 99554491-99554491 on Build GRCh38
rs200904404 Chr.2: 99554438-99554438 on Build GRCh38
rs765114515 Chr.2: 99554441-99554441 on Build GRCh38
rs376932214 Chr.2: 99554451-99554451 on Build GRCh38
rs141110020 Chr.2: 99554396-99554396 on Build GRCh38
rs200443568 Chr.2: 99554420-99554420 on Build GRCh38
rs760098040 Chr.2: 99554457-99554457 on Build GRCh38
rs766875568 Chr.2: 99554401-99554401 on Build GRCh38
rs756690263 Chr.2: 99554455-99554455 on Build GRCh38
rs772657804 Chr.2: 99554486-99554486 on Build GRCh38
rs758389402 Chr.2: 99554448-99554448 on Build GRCh38
rs773374557 Chr.2: 99554407-99554407 on Build GRCh38
rs752486698 Chr.2: 99554446-99554446 on Build GRCh38
rs754085390 Chr.2: 99554435-99554435 on Build GRCh38
rs368703010 Chr.2: 99554449-99554449 on Build GRCh38
rs762394931 Chr.2: 99554399-99554399 on Build GRCh38

View Full Product Details