Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 140776822-140776991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRAF
Entrez Gene ID
673
Gene Name
B-Raf proto-oncogene, serine/threonine kinase
Gene Aliases
B-RAF1, B-raf, BRAF1, NS7, RAFB1
UniGene
Hs.550061
Gene Chromosome Location
Chr.7: 140719331-140924764 on Build GRCh38
Transcripts
NM_004333.4 XM_005250045.2 XM_017012558.1 XM_017012559.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5872003 c.1691T>C p.M564T Chr.7: 140776915-140776915 on Build GRCh38
6943287 c.1615C>T p.H539Y Chr.7: 140776991-140776991 on Build GRCh38
6962126 c.1661T>C p.I554T Chr.7: 140776945-140776945 on Build GRCh38
1167936 c.1624C>T p.H542Y Chr.7: 140776982-140776982 on Build GRCh38
6956195 c.1673G>A p.R558Q Chr.7: 140776933-140776933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781213584 Chr.7: 140776909-140776909 on Build GRCh38
rs756413322 Chr.7: 140776882-140776882 on Build GRCh38
rs960852908 Chr.7: 140776859-140776859 on Build GRCh38
rs768303459 Chr.7: 140776964-140776964 on Build GRCh38
rs368578780 Chr.7: 140776899-140776899 on Build GRCh38
rs760524720 Chr.7: 140776939-140776939 on Build GRCh38
rs766753485 Chr.7: 140776885-140776885 on Build GRCh38
rs1057517930 Chr.7: 140776910-140776910 on Build GRCh38
rs969439754 Chr.7: 140776860-140776860 on Build GRCh38
rs761334141 Chr.7: 140776866-140776866 on Build GRCh38
rs779055779 Chr.7: 140776891-140776891 on Build GRCh38
rs775147056 Chr.7: 140776852-140776852 on Build GRCh38
rs547857147 Chr.7: 140776886-140776886 on Build GRCh38
rs878854675 Chr.7: 140776945-140776945 on Build GRCh38
rs745783052 Chr.7: 140776916-140776916 on Build GRCh38
rs749083550 Chr.7: 140776949-140776949 on Build GRCh38
rs184144181 Chr.7: 140776898-140776898 on Build GRCh38
rs769683898 Chr.7: 140776938-140776938 on Build GRCh38
rs750933612 Chr.7: 140776867-140776867 on Build GRCh38

View Full Product Details