Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48791980-48792152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATA1
Entrez Gene ID
2623
Gene Name
GATA binding protein 1
Gene Aliases
ERYF1, GATA-1, GF-1, GF1, NF-E1, NFE1, XLANP, XLTDA, XLTT
UniGene
Hs.765
Gene Chromosome Location
Chr.X: 48786540-48794311 on Build GRCh38
Transcripts
NM_002049.3 XM_011543897.2 XM_011543898.2

SNP Detail

SNP ID Chromosome Location
rs782265153 Chr.X: 48792122-48792122 on Build GRCh38
rs59609788 Chr.X: 48792102-48792102 on Build GRCh38
rs997764249 Chr.X: 48792042-48792042 on Build GRCh38
rs781852903 Chr.X: 48792021-48792021 on Build GRCh38
rs143332634 Chr.X: 48792103-48792103 on Build GRCh38
rs782651118 Chr.X: 48792105-48792105 on Build GRCh38
rs782527464 Chr.X: 48792025-48792025 on Build GRCh38
rs782588112 Chr.X: 48792073-48792073 on Build GRCh38
rs781904528 Chr.X: 48792041-48792041 on Build GRCh38
rs782756928 Chr.X: 48792016-48792016 on Build GRCh38
rs782788437 Chr.X: 48792029-48792029 on Build GRCh38
rs148357840 Chr.X: 48792124-48792124 on Build GRCh38
rs1050669396 Chr.X: 48792085-48792085 on Build GRCh38
rs782443272 Chr.X: 48792055-48792055 on Build GRCh38

View Full Product Details