Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 13900641-13900815 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETV1
Entrez Gene ID
2115
Gene Name
ETS variant 1
Gene Aliases
ER81
UniGene
Hs.22634
Gene Chromosome Location
Chr.7: 13891229-13991425 on Build GRCh38
Transcripts
NM_001163147.1 NM_001163148.1 NM_001163149.1 NM_001163150.1 NM_001163151.1 NM_001163152.1 NM_004956.4 NR_120445.1 XM_005249635.4 XM_005249636.4 XM_011515167.2 XM_011515168.2 XM_011515170.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6980551 c.1212+1G>A p.? Chr.7: 13900737-13900737 on Build GRCh38
5026704 c.1172G>A p.R391H Chr.7: 13900778-13900778 on Build GRCh38
269120 c.1148C>T p.A383V Chr.7: 13900802-13900802 on Build GRCh38
6054929 c.1212G>A p.K404K Chr.7: 13900738-13900738 on Build GRCh38
5634401 c.1060C>T p.R354C Chr.7: 13900770-13900770 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1026625908 Chr.7: 13900677-13900677 on Build GRCh38
rs766240927 Chr.7: 13900723-13900723 on Build GRCh38
rs951165812 Chr.7: 13900706-13900706 on Build GRCh38
rs79042379 Chr.7: 13900669-13900669 on Build GRCh38
rs773278653 Chr.7: 13900769-13900769 on Build GRCh38
rs775186104 Chr.7: 13900750-13900750 on Build GRCh38
rs564604931 Chr.7: 13900718-13900718 on Build GRCh38
rs748376193 Chr.7: 13900765-13900765 on Build GRCh38
rs772236722 Chr.7: 13900768-13900768 on Build GRCh38
rs757194117 Chr.7: 13900719-13900719 on Build GRCh38
rs752428428 Chr.7: 13900708-13900708 on Build GRCh38
rs760752049 Chr.7: 13900774-13900774 on Build GRCh38
rs369197571 Chr.7: 13900698-13900698 on Build GRCh38
rs3779117 Chr.7: 13900666-13900666 on Build GRCh38
rs180687180 Chr.7: 13900721-13900721 on Build GRCh38
rs758026764 Chr.7: 13900712-13900712 on Build GRCh38
rs778383537 Chr.7: 13900701-13900701 on Build GRCh38
rs541646270 Chr.7: 13900671-13900671 on Build GRCh38
rs777316660 Chr.7: 13900713-13900713 on Build GRCh38
rs373175151 Chr.7: 13900744-13900744 on Build GRCh38
rs1007679355 Chr.7: 13900770-13900770 on Build GRCh38
rs745607441 Chr.7: 13900723-13900723 on Build GRCh38

View Full Product Details