Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 142863601-142863733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHB6
Entrez Gene ID
2051
Gene Name
EPH receptor B6
Gene Aliases
HEP
UniGene
Hs.380089
Gene Chromosome Location
Chr.7: 142854640-142871093 on Build GRCh38
Transcripts
NM_001280794.2 NM_001280795.2 NM_004445.5 NR_104001.2 XM_006715881.2 XM_011515879.1 XM_011515880.1 XM_011515881.2 XM_011515882.2

SNP Detail

SNP ID Chromosome Location
rs373096667 Chr.7: 142863693-142863693 on Build GRCh38
rs747647138 Chr.7: 142863656-142863656 on Build GRCh38
rs772063073 Chr.7: 142863707-142863707 on Build GRCh38
rs773024348 Chr.7: 142863690-142863690 on Build GRCh38
rs763119570 Chr.7: 142863695-142863695 on Build GRCh38
rs533653752 Chr.7: 142863691-142863691 on Build GRCh38
rs758973552 Chr.7: 142863680-142863680 on Build GRCh38
rs753392889 Chr.7: 142863638-142863638 on Build GRCh38
rs200656818 Chr.7: 142863650-142863650 on Build GRCh38
rs748060252 Chr.7: 142863689-142863689 on Build GRCh38
rs778072644 Chr.7: 142863689-142863689 on Build GRCh38
rs771807343 Chr.7: 142863689-142863690 on Build GRCh38
rs747410220 Chr.7: 142863690-142863690 on Build GRCh38
rs754050538 Chr.7: 142863651-142863651 on Build GRCh38

View Full Product Details