Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 42287623-42287796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIC
Entrez Gene ID
23152
Gene Name
capicua transcriptional repressor
Gene Aliases
-
UniGene
Hs.388236
Gene Chromosome Location
Chr.19: 42268537-42295796 on Build GRCh38
Transcripts
NM_001304815.1 NM_015125.4 XM_005258673.2 XM_005258674.2 XM_005258675.2 XM_011526660.2 XM_011526661.2 XM_011526662.2 XM_011526663.2 XM_011526664.2 XM_011526665.2 XM_011526666.2 XM_011526667.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4420727 c.714G>A p.W238* Chr.19: 42287676-42287676 on Build GRCh38
5731279 c.692G>A p.S231N Chr.19: 42287654-42287654 on Build GRCh38
304189 c.699_701delCCT p.I233_L234>M Chr.19: 42287661-42287663 on Build GRCh38
5855937 c.765+1G>A p.? Chr.19: 42287728-42287728 on Build GRCh38
6210443 c.765+3A>T p.? Chr.19: 42287730-42287730 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746928420 Chr.19: 42287686-42287686 on Build GRCh38
rs371737496 Chr.19: 42287696-42287696 on Build GRCh38
rs756026733 Chr.19: 42287758-42287760 on Build GRCh38
rs765094541 Chr.19: 42287706-42287706 on Build GRCh38
rs931970286 Chr.19: 42287682-42287682 on Build GRCh38
rs965861102 Chr.19: 42287762-42287762 on Build GRCh38
rs376425349 Chr.19: 42287732-42287732 on Build GRCh38
rs750378809 Chr.19: 42287744-42287745 on Build GRCh38
rs202213460 Chr.19: 42287652-42287652 on Build GRCh38
rs369225136 Chr.19: 42287770-42287770 on Build GRCh38
rs768135574 Chr.19: 42287758-42287758 on Build GRCh38
rs1020337272 Chr.19: 42287771-42287771 on Build GRCh38
rs753110946 Chr.19: 42287760-42287760 on Build GRCh38
rs773618772 Chr.19: 42287712-42287712 on Build GRCh38
rs764367333 Chr.19: 42287743-42287743 on Build GRCh38
rs185820178 Chr.19: 42287735-42287735 on Build GRCh38
rs774507373 Chr.19: 42287754-42287754 on Build GRCh38
rs567197549 Chr.19: 42287769-42287769 on Build GRCh38
rs756517491 Chr.19: 42287768-42287768 on Build GRCh38
rs895050666 Chr.19: 42287724-42287724 on Build GRCh38
rs1050416819 Chr.19: 42287693-42287693 on Build GRCh38
rs751599325 Chr.19: 42287740-42287740 on Build GRCh38
rs181552919 Chr.19: 42287649-42287649 on Build GRCh38
rs776422599 Chr.19: 42287694-42287694 on Build GRCh38
rs1011591601 Chr.19: 42287761-42287761 on Build GRCh38
rs766598474 Chr.19: 42287733-42287733 on Build GRCh38
rs768583610 Chr.19: 42287690-42287690 on Build GRCh38

View Full Product Details