Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 125629245-125629379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHEK1
Entrez Gene ID
1111
Gene Name
checkpoint kinase 1
Gene Aliases
CHK1
UniGene
Hs.24529
Gene Chromosome Location
Chr.11: 125624910-125676259 on Build GRCh38
Transcripts
NM_001114121.2 NM_001114122.2 NM_001244846.1 NM_001274.5 NR_045204.1 NR_045205.1 XM_011542560.2 XM_011542561.2 XM_011542562.2 XM_011542563.2 XM_017017146.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6268365 c.354+1G>T p.? Chr.11: 125629297-125629297 on Build GRCh38
5768634 c.323A>G p.Q108R Chr.11: 125629265-125629265 on Build GRCh38
4019053 c.351G>T p.G117G Chr.11: 125629293-125629293 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs201570663 Chr.11: 125629350-125629350 on Build GRCh38
rs747265349 Chr.11: 125629348-125629348 on Build GRCh38
rs754651841 Chr.11: 125629300-125629300 on Build GRCh38
rs766064275 Chr.11: 125629297-125629297 on Build GRCh38
rs375305289 Chr.11: 125629296-125629296 on Build GRCh38
rs777648224 Chr.11: 125629331-125629331 on Build GRCh38
rs752465693 Chr.11: 125629313-125629313 on Build GRCh38
rs201903974 Chr.11: 125629318-125629318 on Build GRCh38
rs780329343 Chr.11: 125629345-125629345 on Build GRCh38
rs1048249612 Chr.11: 125629291-125629291 on Build GRCh38
rs745970536 Chr.11: 125629337-125629337 on Build GRCh38
rs777189541 Chr.11: 125629351-125629351 on Build GRCh38
rs751102775 Chr.11: 125629298-125629298 on Build GRCh38
rs781020833 Chr.11: 125629302-125629302 on Build GRCh38
rs867318349 Chr.11: 125629275-125629275 on Build GRCh38
rs771989402 Chr.11: 125629341-125629341 on Build GRCh38
rs563722813 Chr.11: 125629273-125629273 on Build GRCh38
rs749996823 Chr.11: 125629295-125629295 on Build GRCh38

View Full Product Details