Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 8058396-8058570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATA3
Entrez Gene ID
2625
Gene Name
GATA binding protein 3
Gene Aliases
HDR, HDRS
UniGene
Hs.524134
Gene Chromosome Location
Chr.10: 8045421-8075201 on Build GRCh38
Transcripts
NM_001002295.1 NM_002051.2 XM_005252442.2 XM_005252443.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6943176 c.400C>T p.P134S Chr.10: 8058463-8058463 on Build GRCh38
920712 c.488C>T p.P163L Chr.10: 8058551-8058551 on Build GRCh38
6981089 c.403C>A p.P135T Chr.10: 8058466-8058466 on Build GRCh38
4774998 c.492_493insT p.P165fs*139 Chr.10: 8058555-8058556 on Build GRCh38
5811774 c.401C>A p.P134H Chr.10: 8058464-8058464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750735308 Chr.10: 8058422-8058422 on Build GRCh38
rs143627754 Chr.10: 8058543-8058543 on Build GRCh38
rs746851056 Chr.10: 8058465-8058465 on Build GRCh38
rs547425013 Chr.10: 8058434-8058435 on Build GRCh38
rs772396478 Chr.10: 8058461-8058462 on Build GRCh38
rs772388081 Chr.10: 8058462-8058462 on Build GRCh38
rs201023943 Chr.10: 8058551-8058551 on Build GRCh38
rs756446087 Chr.10: 8058427-8058427 on Build GRCh38
rs773322691 Chr.10: 8058472-8058474 on Build GRCh38
rs757135223 Chr.10: 8058519-8058519 on Build GRCh38
rs778182252 Chr.10: 8058508-8058508 on Build GRCh38
rs777735536 Chr.10: 8058464-8058464 on Build GRCh38
rs991194737 Chr.10: 8058457-8058457 on Build GRCh38
rs748390189 Chr.10: 8058444-8058444 on Build GRCh38
rs780782586 Chr.10: 8058523-8058523 on Build GRCh38
rs587778379 Chr.10: 8058511-8058511 on Build GRCh38
rs774820359 Chr.10: 8058475-8058475 on Build GRCh38
rs770601456 Chr.10: 8058467-8058467 on Build GRCh38
rs36100495 Chr.10: 8058487-8058488 on Build GRCh38
rs755377699 Chr.10: 8058495-8058495 on Build GRCh38
rs142451494 Chr.10: 8058438-8058438 on Build GRCh38
rs745734873 Chr.10: 8058474-8058474 on Build GRCh38
rs758531534 Chr.10: 8058507-8058507 on Build GRCh38
rs1016713478 Chr.10: 8058537-8058537 on Build GRCh38
rs769567541 Chr.10: 8058528-8058528 on Build GRCh38
rs150906764 Chr.10: 8058466-8058466 on Build GRCh38
rs969928437 Chr.10: 8058510-8058510 on Build GRCh38
rs200317918 Chr.10: 8058489-8058489 on Build GRCh38
rs745848216 Chr.10: 8058526-8058526 on Build GRCh38
rs766774125 Chr.10: 8058552-8058552 on Build GRCh38
rs373234853 Chr.10: 8058497-8058497 on Build GRCh38
rs748832905 Chr.10: 8058534-8058534 on Build GRCh38
rs766636397 Chr.10: 8058491-8058491 on Build GRCh38
rs775381986 Chr.10: 8058531-8058531 on Build GRCh38
rs575374217 Chr.10: 8058484-8058484 on Build GRCh38
rs561866523 Chr.10: 8058546-8058546 on Build GRCh38
rs765392255 Chr.10: 8058498-8058498 on Build GRCh38
rs776699046 Chr.10: 8058468-8058468 on Build GRCh38
rs992348054 Chr.10: 8058529-8058529 on Build GRCh38
rs545553575 Chr.10: 8058486-8058486 on Build GRCh38
rs535492085 Chr.10: 8058436-8058436 on Build GRCh38
rs959805514 Chr.10: 8058456-8058456 on Build GRCh38
rs754386700 Chr.10: 8058494-8058494 on Build GRCh38
rs752779081 Chr.10: 8058499-8058499 on Build GRCh38
rs780648121 Chr.10: 8058435-8058435 on Build GRCh38

View Full Product Details