Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52403089-52403236 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAP1
Entrez Gene ID
8314
Gene Name
BRCA1 associated protein 1
Gene Aliases
HUCEP-13, UCHL2, hucep-6
UniGene
Hs.106674
Gene Chromosome Location
Chr.3: 52401004-52410105 on Build GRCh38
Transcripts
NM_004656.3 XM_011534149.2 XM_011534150.2 XM_011534151.2 XM_011534152.1 XM_017007303.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1738879 c.1825A>G p.S609G Chr.3: 52403203-52403203 on Build GRCh38
6909679 c.1807_1829del23 p.V603fs*32 Chr.3: 52403199-52403221 on Build GRCh38
6980158 c.1829G>A p.R610K Chr.3: 52403199-52403199 on Build GRCh38
6922636 c.1840_1851>T p.G614fs*25 Chr.3: 52403177-52403188 on Build GRCh38
6023803 c.1869_1872delTGGG p.S623fs*13 Chr.3: 52403156-52403159 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866649101 Chr.3: 52403167-52403167 on Build GRCh38
rs537090290 Chr.3: 52403127-52403127 on Build GRCh38
rs758585834 Chr.3: 52403170-52403170 on Build GRCh38
rs964152039 Chr.3: 52403147-52403147 on Build GRCh38
rs752063900 Chr.3: 52403207-52403207 on Build GRCh38
rs375950004 Chr.3: 52403156-52403156 on Build GRCh38
rs752636233 Chr.3: 52403165-52403165 on Build GRCh38
rs371236827 Chr.3: 52403114-52403114 on Build GRCh38
rs35448940 Chr.3: 52403190-52403190 on Build GRCh38
rs989620105 Chr.3: 52403118-52403118 on Build GRCh38
rs749728488 Chr.3: 52403191-52403191 on Build GRCh38
rs781269263 Chr.3: 52403186-52403186 on Build GRCh38
rs776553452 Chr.3: 52403128-52403128 on Build GRCh38
rs746271680 Chr.3: 52403188-52403188 on Build GRCh38
rs772835408 Chr.3: 52403201-52403201 on Build GRCh38
rs751746876 Chr.3: 52403178-52403178 on Build GRCh38
rs757559034 Chr.3: 52403180-52403180 on Build GRCh38
rs756450018 Chr.3: 52403189-52403189 on Build GRCh38
rs759370336 Chr.3: 52403132-52403132 on Build GRCh38
rs770654975 Chr.3: 52403208-52403208 on Build GRCh38
rs771494475 Chr.3: 52403195-52403195 on Build GRCh38
rs35353781 Chr.3: 52403181-52403181 on Build GRCh38
rs116604051 Chr.3: 52403171-52403171 on Build GRCh38

View Full Product Details