Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 77815593-77815765 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAQ
Entrez Gene ID
2776
Gene Name
G protein subunit alpha q
Gene Aliases
CMC1, G-ALPHA-q, GAQ, SWS
UniGene
Hs.269782
Gene Chromosome Location
Chr.9: 77716274-78031449 on Build GRCh38
Transcripts
NM_002072.4 XM_017014628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6116259 c.436G>C p.D146H Chr.9: 77815656-77815656 on Build GRCh38
6367443 c.281C>T p.A94V Chr.9: 77815724-77815724 on Build GRCh38
5083709 c.397_400delAAGA p.K133fs*34 Chr.9: 77815692-77815695 on Build GRCh38
6365064 c.382A>G p.T128A Chr.9: 77815623-77815623 on Build GRCh38
1110327 c.340C>T p.R114* Chr.9: 77815752-77815752 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770370257 Chr.9: 77815738-77815738 on Build GRCh38
rs750558344 Chr.9: 77815663-77815663 on Build GRCh38
rs145171922 Chr.9: 77815711-77815711 on Build GRCh38
rs758684158 Chr.9: 77815672-77815672 on Build GRCh38
rs923027293 Chr.9: 77815623-77815623 on Build GRCh38
rs779143139 Chr.9: 77815640-77815640 on Build GRCh38
rs748772794 Chr.9: 77815729-77815729 on Build GRCh38
rs368906365 Chr.9: 77815690-77815690 on Build GRCh38
rs34939122 Chr.9: 77815672-77815673 on Build GRCh38
rs748011987 Chr.9: 77815691-77815691 on Build GRCh38
rs192927818 Chr.9: 77815704-77815704 on Build GRCh38
rs200924131 Chr.9: 77815636-77815636 on Build GRCh38

View Full Product Details