Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89739051-89739217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCA
Entrez Gene ID
2175
Gene Name
Fanconi anemia complementation group A
Gene Aliases
FA, FA-H, FA1, FAA, FACA, FAH, FANCH
UniGene
Hs.744083
Gene Chromosome Location
Chr.16: 89737551-89816658 on Build GRCh38
Transcripts
NM_000135.2 NM_001286167.1 XM_005256294.4 XM_011522945.2 XM_011522946.2 XM_011522947.2 XM_017023044.1 NM_001113525.1 NM_152287.3 NR_110122.1 NR_110126.1 NR_110128.1 NR_110129.1 XM_017023890.1

Gene Symbol
ZNF276
Entrez Gene ID
92822
Gene Name
zinc finger protein 276
Gene Aliases
CENP-Z, CENPZ, ZADT, ZFP276, ZNF477
UniGene
Hs.290154
Gene Chromosome Location
Chr.16: 89720368-89740925 on Build GRCh38
Transcripts
NM_000135.2 NM_001286167.1 XM_005256294.4 XM_011522945.2 XM_011522946.2 XM_011522947.2 XM_017023044.1 NM_001113525.1 NM_152287.3 NR_110122.1 NR_110126.1 NR_110128.1 NR_110129.1 XM_017023890.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6953813 c.4114G>A p.D1372N Chr.16: 89739186-89739186 on Build GRCh38
4129524 c.4115A>G p.D1372G Chr.16: 89739185-89739185 on Build GRCh38
5575680 c.4097A>G p.Q1366R Chr.16: 89739203-89739203 on Build GRCh38
6918716 c.4098G>T p.Q1366H Chr.16: 89739202-89739202 on Build GRCh38
3513342 c.4146G>A p.R1382R Chr.16: 89739154-89739154 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372493612 Chr.16: 89739165-89739165 on Build GRCh38
rs779568987 Chr.16: 89739089-89739089 on Build GRCh38
rs367970303 Chr.16: 89739155-89739155 on Build GRCh38
rs774175580 Chr.16: 89739152-89739152 on Build GRCh38
rs377636161 Chr.16: 89739090-89739090 on Build GRCh38
rs752700058 Chr.16: 89739083-89739083 on Build GRCh38
rs757550124 Chr.16: 89739110-89739110 on Build GRCh38
rs1005560269 Chr.16: 89739191-89739191 on Build GRCh38
rs776244734 Chr.16: 89739091-89739091 on Build GRCh38
rs746203082 Chr.16: 89739112-89739112 on Build GRCh38
rs759521427 Chr.16: 89739096-89739096 on Build GRCh38
rs769115368 Chr.16: 89739137-89739137 on Build GRCh38
rs769505201 Chr.16: 89739092-89739092 on Build GRCh38
rs757592625 Chr.16: 89739189-89739189 on Build GRCh38
rs531372521 Chr.16: 89739143-89739143 on Build GRCh38
rs768765651 Chr.16: 89739098-89739099 on Build GRCh38
rs199599393 Chr.16: 89739170-89739170 on Build GRCh38
rs371366647 Chr.16: 89739106-89739106 on Build GRCh38
rs553955430 Chr.16: 89739078-89739078 on Build GRCh38
rs775234845 Chr.16: 89739162-89739162 on Build GRCh38
rs201225325 Chr.16: 89739195-89739195 on Build GRCh38
rs766226560 Chr.16: 89739093-89739093 on Build GRCh38
rs374765708 Chr.16: 89739131-89739131 on Build GRCh38
rs375237681 Chr.16: 89739196-89739196 on Build GRCh38
rs747194256 Chr.16: 89739090-89739091 on Build GRCh38
rs781393563 Chr.16: 89739081-89739081 on Build GRCh38
rs759216137 Chr.16: 89739154-89739154 on Build GRCh38
rs191404781 Chr.16: 89739087-89739087 on Build GRCh38
rs751344774 Chr.16: 89739094-89739094 on Build GRCh38
rs779466859 Chr.16: 89739111-89739111 on Build GRCh38
rs754048546 Chr.16: 89739109-89739109 on Build GRCh38
rs772665586 Chr.16: 89739125-89739125 on Build GRCh38
rs772701240 Chr.16: 89739138-89739138 on Build GRCh38
rs776969626 Chr.16: 89739175-89739176 on Build GRCh38
rs750839091 Chr.16: 89739192-89739192 on Build GRCh38
rs200304203 Chr.16: 89739107-89739107 on Build GRCh38
rs753944130 Chr.16: 89739183-89739183 on Build GRCh38
rs370080063 Chr.16: 89739129-89739129 on Build GRCh38
rs780301799 Chr.16: 89739086-89739087 on Build GRCh38
rs760814763 Chr.16: 89739167-89739167 on Build GRCh38
rs765515655 Chr.16: 89739190-89739190 on Build GRCh38
rs756403983 Chr.16: 89739086-89739086 on Build GRCh38
rs770551622 Chr.16: 89739145-89739145 on Build GRCh38
rs545928949 Chr.16: 89739104-89739104 on Build GRCh38

View Full Product Details