Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 41225082-41225240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNNB1
Entrez Gene ID
1499
Gene Name
catenin beta 1
Gene Aliases
CTNNB, MRD19, armadillo
UniGene
Hs.476018
Gene Chromosome Location
Chr.3: 41199451-41240448 on Build GRCh38
Transcripts
NM_001098209.1 NM_001098210.1 NM_001904.3 XM_005264886.2 XM_006712983.1 XM_006712984.1 XM_006712985.1 XM_017005738.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6941850 c.102_416del515 p.? Chr.3: 41224614-41225128 on Build GRCh38
6035 c.14_430del417 p.A5_Q143del Chr.3: 41224526-41225142 on Build GRCh38
446497 c.390G>A p.Q130Q Chr.3: 41225102-41225102 on Build GRCh38
396036 c.419T>C p.I140T Chr.3: 41225131-41225131 on Build GRCh38
6033 c.14_424del411 p.A5_Y142>D Chr.3: 41224526-41225136 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369510063 Chr.3: 41225195-41225195 on Build GRCh38
rs778138109 Chr.3: 41225108-41225108 on Build GRCh38
rs749833775 Chr.3: 41225129-41225129 on Build GRCh38
rs200968230 Chr.3: 41225164-41225164 on Build GRCh38
rs775491694 Chr.3: 41225106-41225106 on Build GRCh38
rs5743392 Chr.3: 41225198-41225198 on Build GRCh38
rs774085540 Chr.3: 41225174-41225174 on Build GRCh38
rs771554085 Chr.3: 41225187-41225187 on Build GRCh38
rs892322076 Chr.3: 41225148-41225148 on Build GRCh38
rs745411917 Chr.3: 41225180-41225180 on Build GRCh38
rs3856746 Chr.3: 41225132-41225132 on Build GRCh38

View Full Product Details