Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45352236-45352397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC2
Entrez Gene ID
2068
Gene Name
ERCC excision repair 2, TFIIH core complex helicase subunit
Gene Aliases
COFS2, EM9, TFIIH, TTD, TTD1, XPD
UniGene
Hs.487294
Gene Chromosome Location
Chr.19: 45349837-45370647 on Build GRCh38
Transcripts
NM_000400.3 XM_011526611.2 XM_017026467.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3104029 c.2068C>T p.R690W Chr.19: 45352331-45352331 on Build GRCh38
4950918 c.2057G>C p.R686P Chr.19: 45352342-45352342 on Build GRCh38
232265 c.2131G>T p.D711Y Chr.19: 45352268-45352268 on Build GRCh38
7323630 c.2137G>A p.G713S Chr.19: 45352262-45352262 on Build GRCh38
6952893 c.2069G>T p.R690L Chr.19: 45352330-45352330 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201392911 Chr.19: 45352316-45352316 on Build GRCh38
rs778708101 Chr.19: 45352294-45352294 on Build GRCh38
rs750996893 Chr.19: 45352273-45352273 on Build GRCh38
rs886054495 Chr.19: 45352305-45352305 on Build GRCh38
rs775212976 Chr.19: 45352265-45352265 on Build GRCh38
rs999070778 Chr.19: 45352306-45352306 on Build GRCh38
rs772055555 Chr.19: 45352260-45352260 on Build GRCh38
rs537616689 Chr.19: 45352330-45352330 on Build GRCh38
rs747842557 Chr.19: 45352367-45352367 on Build GRCh38
rs780405292 Chr.19: 45352275-45352275 on Build GRCh38
rs758758729 Chr.19: 45352274-45352274 on Build GRCh38
rs768554709 Chr.19: 45352363-45352363 on Build GRCh38
rs374801728 Chr.19: 45352366-45352366 on Build GRCh38
rs756273306 Chr.19: 45352288-45352288 on Build GRCh38
rs141808167 Chr.19: 45352271-45352271 on Build GRCh38
rs771934871 Chr.19: 45352301-45352301 on Build GRCh38
rs930757411 Chr.19: 45352371-45352371 on Build GRCh38
rs144277365 Chr.19: 45352343-45352343 on Build GRCh38
rs546902024 Chr.19: 45352257-45352257 on Build GRCh38
rs751956427 Chr.19: 45352331-45352331 on Build GRCh38
rs763610846 Chr.19: 45352268-45352268 on Build GRCh38
rs373135900 Chr.19: 45352358-45352358 on Build GRCh38
rs41556519 Chr.19: 45352352-45352352 on Build GRCh38
rs746618110 Chr.19: 45352315-45352315 on Build GRCh38
rs147128863 Chr.19: 45352272-45352272 on Build GRCh38
rs1052555 Chr.19: 45352266-45352266 on Build GRCh38
rs761933302 Chr.19: 45352340-45352340 on Build GRCh38
rs121913022 Chr.19: 45352262-45352262 on Build GRCh38
rs999439847 Chr.19: 45352269-45352269 on Build GRCh38
rs775446858 Chr.19: 45352312-45352312 on Build GRCh38
rs140296400 Chr.19: 45352285-45352285 on Build GRCh38
rs745801466 Chr.19: 45352296-45352296 on Build GRCh38
rs774392894 Chr.19: 45352256-45352256 on Build GRCh38
rs758439420 Chr.19: 45352351-45352351 on Build GRCh38
rs745750247 Chr.19: 45352348-45352348 on Build GRCh38
rs763468590 Chr.19: 45352329-45352329 on Build GRCh38
rs764868582 Chr.19: 45352319-45352319 on Build GRCh38
rs755217625 Chr.19: 45352283-45352283 on Build GRCh38
rs751727363 Chr.19: 45352279-45352279 on Build GRCh38
rs773642945 Chr.19: 45352323-45352323 on Build GRCh38
rs776240435 Chr.19: 45352365-45352370 on Build GRCh38
rs557601608 Chr.19: 45352342-45352342 on Build GRCh38
rs777919686 Chr.19: 45352344-45352344 on Build GRCh38

View Full Product Details