Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 187724819-187724984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL6
Entrez Gene ID
604
Gene Name
B-cell CLL/lymphoma 6
Gene Aliases
BCL5, BCL6A, LAZ3, ZBTB27, ZNF51
UniGene
Hs.478588
Gene Chromosome Location
Chr.3: 187721377-187745725 on Build GRCh38
Transcripts
NM_001130845.1 NM_001134738.1 NM_001706.4 XM_005247694.3 XM_011513062.2 NR_034062.1

Gene Symbol
LOC100131635
Entrez Gene ID
100131635
Gene Name
hCG1645011-like
Gene Aliases
-
UniGene
Hs.434448
Gene Chromosome Location
Chr.3: 187702366-187732415 on Build GRCh38
Transcripts
NM_001130845.1 NM_001134738.1 NM_001706.4 XM_005247694.3 XM_011513062.2 NR_034062.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1731947 c.1977+3A>G p.? Chr.3: 187724938-187724938 on Build GRCh38
1042446 c.1948C>T p.R650* Chr.3: 187724970-187724970 on Build GRCh38
3427441 c.1949G>A p.R650Q Chr.3: 187724969-187724969 on Build GRCh38
5527311 c.1954C>T p.H652Y Chr.3: 187724964-187724964 on Build GRCh38
1235220 c.1940G>A p.S647N Chr.3: 187724978-187724978 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs991461177 Chr.3: 187724868-187724868 on Build GRCh38
rs558189829 Chr.3: 187724919-187724919 on Build GRCh38
rs370413402 Chr.3: 187724922-187724922 on Build GRCh38
rs918152896 Chr.3: 187724871-187724871 on Build GRCh38
rs544868439 Chr.3: 187724916-187724916 on Build GRCh38
rs199744143 Chr.3: 187724874-187724875 on Build GRCh38
rs201578580 Chr.3: 187724931-187724931 on Build GRCh38
rs151236520 Chr.3: 187724962-187724962 on Build GRCh38
rs199791064 Chr.3: 187724854-187724855 on Build GRCh38
rs757780738 Chr.3: 187724924-187724924 on Build GRCh38
rs752841262 Chr.3: 187724912-187724912 on Build GRCh38
rs79388315 Chr.3: 187724874-187724874 on Build GRCh38
rs761695899 Chr.3: 187724893-187724893 on Build GRCh38
rs765176575 Chr.3: 187724900-187724900 on Build GRCh38
rs114017848 Chr.3: 187724873-187724873 on Build GRCh38
rs776048282 Chr.3: 187724938-187724938 on Build GRCh38
rs374762310 Chr.3: 187724925-187724925 on Build GRCh38
rs112100267 Chr.3: 187724937-187724937 on Build GRCh38
rs981003253 Chr.3: 187724848-187724848 on Build GRCh38
rs201148881 Chr.3: 187724920-187724920 on Build GRCh38
rs756180658 Chr.3: 187724934-187724935 on Build GRCh38
rs747647309 Chr.3: 187724947-187724947 on Build GRCh38
rs750747913 Chr.3: 187724855-187724855 on Build GRCh38
rs566972919 Chr.3: 187724854-187724854 on Build GRCh38
rs766240312 Chr.3: 187724906-187724906 on Build GRCh38
rs202094292 Chr.3: 187724905-187724905 on Build GRCh38
rs200408610 Chr.3: 187724926-187724926 on Build GRCh38
rs946034649 Chr.3: 187724908-187724908 on Build GRCh38
rs780513336 Chr.3: 187724932-187724932 on Build GRCh38

View Full Product Details