Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 128481103-128481266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATA2
Entrez Gene ID
2624
Gene Name
GATA binding protein 2
Gene Aliases
DCML, IMD21, MONOMAC, NFE1B
UniGene
Hs.367725
Gene Chromosome Location
Chr.3: 128479422-128493187 on Build GRCh38
Transcripts
NM_001145661.1 NM_001145662.1 NM_032638.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6964801 c.1259G>A p.C420Y Chr.3: 128481203-128481203 on Build GRCh38
1418767 c.1281delC p.F428fs*49 Chr.3: 128481181-128481181 on Build GRCh38
727595 c.1332G>A p.P444P Chr.3: 128481130-128481130 on Build GRCh38
6962786 c.1231G>A p.A411T Chr.3: 128481231-128481231 on Build GRCh38
6669543 c.1255A>G p.K419E Chr.3: 128481207-128481207 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1009940359 Chr.3: 128481137-128481137 on Build GRCh38
rs773731858 Chr.3: 128481156-128481156 on Build GRCh38
rs976064371 Chr.3: 128481128-128481128 on Build GRCh38
rs146554939 Chr.3: 128481188-128481188 on Build GRCh38
rs758517602 Chr.3: 128481195-128481195 on Build GRCh38
rs775080624 Chr.3: 128481142-128481142 on Build GRCh38
rs751577185 Chr.3: 128481220-128481220 on Build GRCh38
rs770768777 Chr.3: 128481130-128481130 on Build GRCh38
rs1037327872 Chr.3: 128481203-128481203 on Build GRCh38
rs984462931 Chr.3: 128481196-128481196 on Build GRCh38
rs764442526 Chr.3: 128481201-128481201 on Build GRCh38
rs886057929 Chr.3: 128481136-128481136 on Build GRCh38
rs768399393 Chr.3: 128481150-128481150 on Build GRCh38
rs761447305 Chr.3: 128481168-128481168 on Build GRCh38
rs201155045 Chr.3: 128481176-128481176 on Build GRCh38
rs909675639 Chr.3: 128481180-128481180 on Build GRCh38
rs747150675 Chr.3: 128481122-128481122 on Build GRCh38
rs971128233 Chr.3: 128481160-128481160 on Build GRCh38
rs777283732 Chr.3: 128481175-128481175 on Build GRCh38
rs766740489 Chr.3: 128481172-128481172 on Build GRCh38
rs745601795 Chr.3: 128481132-128481132 on Build GRCh38
rs781161922 Chr.3: 128481131-128481131 on Build GRCh38
rs374457534 Chr.3: 128481230-128481230 on Build GRCh38
rs1126559 Chr.3: 128481144-128481144 on Build GRCh38
rs762576353 Chr.3: 128481146-128481146 on Build GRCh38
rs769577309 Chr.3: 128481133-128481133 on Build GRCh38
rs753159607 Chr.3: 128481189-128481189 on Build GRCh38
rs34172218 Chr.3: 128481229-128481229 on Build GRCh38

View Full Product Details