Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 241504198-241504337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FH
Entrez Gene ID
2271
Gene Name
fumarate hydratase
Gene Aliases
FMRD, HLRCC, LRCC, MCL, MCUL1
UniGene
Hs.592490
Gene Chromosome Location
Chr.1: 241497557-241519785 on Build GRCh38
Transcripts
NM_000143.3 XM_011544132.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7101140 c.939A>T p.E313D Chr.1: 241504211-241504211 on Build GRCh38
906406 c.942T>C p.A314A Chr.1: 241504208-241504208 on Build GRCh38
5019709 c.927G>A p.P309P Chr.1: 241504223-241504223 on Build GRCh38
6942028 c.925C>T p.P309S Chr.1: 241504225-241504225 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs182531846 Chr.1: 241504266-241504266 on Build GRCh38
rs989876911 Chr.1: 241504237-241504237 on Build GRCh38
rs794727836 Chr.1: 241504232-241504238 on Build GRCh38
rs797044973 Chr.1: 241504246-241504246 on Build GRCh38
rs61737760 Chr.1: 241504223-241504223 on Build GRCh38
rs200491078 Chr.1: 241504239-241504239 on Build GRCh38
rs774312388 Chr.1: 241504296-241504296 on Build GRCh38
rs756528378 Chr.1: 241504224-241504224 on Build GRCh38
rs200809528 Chr.1: 241504284-241504284 on Build GRCh38
rs201502246 Chr.1: 241504242-241504242 on Build GRCh38
rs769561153 Chr.1: 241504282-241504282 on Build GRCh38
rs147991516 Chr.1: 241504233-241504233 on Build GRCh38
rs121913118 Chr.1: 241504228-241504228 on Build GRCh38
rs201168038 Chr.1: 241504293-241504293 on Build GRCh38
rs886046318 Chr.1: 241504250-241504250 on Build GRCh38
rs368849989 Chr.1: 241504225-241504225 on Build GRCh38
rs144597860 Chr.1: 241504290-241504290 on Build GRCh38
rs367887305 Chr.1: 241504275-241504275 on Build GRCh38
rs199539806 Chr.1: 241504273-241504273 on Build GRCh38
rs746479591 Chr.1: 241504263-241504263 on Build GRCh38
rs745515367 Chr.1: 241504232-241504232 on Build GRCh38
rs553516187 Chr.1: 241504289-241504289 on Build GRCh38

View Full Product Details