Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 187722432-187722606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL6
Entrez Gene ID
604
Gene Name
B-cell CLL/lymphoma 6
Gene Aliases
BCL5, BCL6A, LAZ3, ZBTB27, ZNF51
UniGene
Hs.478588
Gene Chromosome Location
Chr.3: 187721377-187745725 on Build GRCh38
Transcripts
NM_001130845.1 NM_001134738.1 NM_001706.4 XM_005247694.3 XM_011513062.2 NR_034062.1

Gene Symbol
LOC100131635
Entrez Gene ID
100131635
Gene Name
hCG1645011-like
Gene Aliases
-
UniGene
Hs.434448
Gene Chromosome Location
Chr.3: 187702366-187732415 on Build GRCh38
Transcripts
NM_001130845.1 NM_001134738.1 NM_001706.4 XM_005247694.3 XM_011513062.2 NR_034062.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6982663 c.2064G>T p.K688N Chr.3: 187722515-187722515 on Build GRCh38
5989968 c.2060C>G p.T687S Chr.3: 187722519-187722519 on Build GRCh38
5642270 c.2038A>G p.K680E Chr.3: 187722541-187722541 on Build GRCh38
1042445 c.2033G>A p.R678H Chr.3: 187722546-187722546 on Build GRCh38
6841348 c.2003G>A p.R668H Chr.3: 187722576-187722576 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755174150 Chr.3: 187722505-187722505 on Build GRCh38
rs760088914 Chr.3: 187722480-187722480 on Build GRCh38
rs764958840 Chr.3: 187722503-187722503 on Build GRCh38
rs1056936 Chr.3: 187722553-187722553 on Build GRCh38
rs775007455 Chr.3: 187722479-187722479 on Build GRCh38
rs750212558 Chr.3: 187722504-187722504 on Build GRCh38
rs771401698 Chr.3: 187722454-187722454 on Build GRCh38
rs1003186327 Chr.3: 187722487-187722487 on Build GRCh38
rs888025733 Chr.3: 187722512-187722512 on Build GRCh38
rs1044743980 Chr.3: 187722455-187722455 on Build GRCh38
rs771627344 Chr.3: 187722465-187722465 on Build GRCh38
rs200887028 Chr.3: 187722502-187722502 on Build GRCh38
rs759188830 Chr.3: 187722464-187722464 on Build GRCh38
rs200467709 Chr.3: 187722542-187722542 on Build GRCh38
rs774878142 Chr.3: 187722456-187722456 on Build GRCh38
rs142355235 Chr.3: 187722533-187722533 on Build GRCh38
rs558388208 Chr.3: 187722529-187722529 on Build GRCh38
rs753741309 Chr.3: 187722499-187722499 on Build GRCh38
rs146998695 Chr.3: 187722497-187722497 on Build GRCh38
rs756234161 Chr.3: 187722539-187722539 on Build GRCh38

View Full Product Details