Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41879508-41879682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD79A
Entrez Gene ID
973
Gene Name
CD79a molecule
Gene Aliases
IGA, MB-1
UniGene
Hs.631567
Gene Chromosome Location
Chr.19: 41877120-41881372 on Build GRCh38
Transcripts
NM_001783.3 NM_021601.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7349119 c.395C>T p.P132L Chr.19: 41879550-41879550 on Build GRCh38
6961052 c.408G>A p.M136I Chr.19: 41879563-41879563 on Build GRCh38
6905979 c.445G>A p.G149R Chr.19: 41879600-41879600 on Build GRCh38
4549711 c.480G>A p.G160G Chr.19: 41879635-41879635 on Build GRCh38
7326684 c.411G>A p.G137G Chr.19: 41879566-41879566 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781984117 Chr.19: 41879638-41879638 on Build GRCh38
rs782079466 Chr.19: 41879551-41879551 on Build GRCh38
rs201206281 Chr.19: 41879550-41879550 on Build GRCh38
rs782150073 Chr.19: 41879621-41879621 on Build GRCh38
rs758922246 Chr.19: 41879567-41879567 on Build GRCh38
rs148797987 Chr.19: 41879574-41879574 on Build GRCh38
rs782128528 Chr.19: 41879557-41879557 on Build GRCh38
rs782414774 Chr.19: 41879637-41879637 on Build GRCh38
rs141601187 Chr.19: 41879599-41879599 on Build GRCh38
rs782338772 Chr.19: 41879663-41879663 on Build GRCh38
rs11539391 Chr.19: 41879598-41879598 on Build GRCh38
rs782244248 Chr.19: 41879648-41879648 on Build GRCh38
rs782307227 Chr.19: 41879538-41879538 on Build GRCh38
rs782653837 Chr.19: 41879615-41879615 on Build GRCh38
rs782014534 Chr.19: 41879539-41879539 on Build GRCh38
rs782741265 Chr.19: 41879563-41879563 on Build GRCh38
rs782123960 Chr.19: 41879644-41879644 on Build GRCh38
rs782159652 Chr.19: 41879543-41879543 on Build GRCh38
rs782517457 Chr.19: 41879611-41879611 on Build GRCh38
rs782751776 Chr.19: 41879549-41879549 on Build GRCh38
rs782765449 Chr.19: 41879623-41879623 on Build GRCh38
rs537675914 Chr.19: 41879620-41879620 on Build GRCh38
rs781783087 Chr.19: 41879569-41879569 on Build GRCh38
rs202097187 Chr.19: 41879622-41879622 on Build GRCh38

View Full Product Details