Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 64189956-64190127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMER1
Entrez Gene ID
139285
Gene Name
APC membrane recruitment protein 1
Gene Aliases
FAM123B, OSCS, WTX
UniGene
Hs.314225
Gene Chromosome Location
Chr.X: 64185117-64205744 on Build GRCh38
Transcripts
NM_152424.3

SNP Detail

SNP ID Chromosome Location
rs773261453 Chr.X: 64190080-64190080 on Build GRCh38
rs56253937 Chr.X: 64190033-64190033 on Build GRCh38
rs761842441 Chr.X: 64190015-64190015 on Build GRCh38
rs1043065176 Chr.X: 64190059-64190059 on Build GRCh38
rs761082647 Chr.X: 64190096-64190096 on Build GRCh38
rs773592043 Chr.X: 64190075-64190075 on Build GRCh38
rs766861490 Chr.X: 64190103-64190103 on Build GRCh38
rs200157619 Chr.X: 64190014-64190014 on Build GRCh38
rs768536859 Chr.X: 64189998-64189998 on Build GRCh38
rs749264086 Chr.X: 64189983-64189983 on Build GRCh38
rs754211727 Chr.X: 64190106-64190106 on Build GRCh38
rs375850061 Chr.X: 64190016-64190016 on Build GRCh38

View Full Product Details