Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 77797411-77797561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAQ
Entrez Gene ID
2776
Gene Name
G protein subunit alpha q
Gene Aliases
CMC1, G-ALPHA-q, GAQ, SWS
UniGene
Hs.269782
Gene Chromosome Location
Chr.9: 77716274-78031449 on Build GRCh38
Transcripts
NM_002072.4 XM_017014628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5608951 c.579C>T p.P193P Chr.9: 77797546-77797546 on Build GRCh38
5608952 c.492C>T p.P164P Chr.9: 77797546-77797546 on Build GRCh38
1463116 c.576C>T p.Y192Y Chr.9: 77797549-77797549 on Build GRCh38
6342236 c.571G>A p.E191K Chr.9: 77797554-77797554 on Build GRCh38
4948197 c.489C>T p.Y163Y Chr.9: 77797549-77797549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748225408 Chr.9: 77797504-77797504 on Build GRCh38
rs772790576 Chr.9: 77797475-77797485 on Build GRCh38
rs372125818 Chr.9: 77797498-77797498 on Build GRCh38
rs950025124 Chr.9: 77797437-77797437 on Build GRCh38
rs760448781 Chr.9: 77797470-77797470 on Build GRCh38
rs1041707248 Chr.9: 77797444-77797444 on Build GRCh38
rs547672512 Chr.9: 77797464-77797464 on Build GRCh38
rs969519565 Chr.9: 77797508-77797508 on Build GRCh38
rs375792308 Chr.9: 77797495-77797495 on Build GRCh38
rs935284047 Chr.9: 77797455-77797455 on Build GRCh38
rs369912964 Chr.9: 77797496-77797496 on Build GRCh38
rs761352239 Chr.9: 77797494-77797494 on Build GRCh38
rs376025257 Chr.9: 77797448-77797448 on Build GRCh38
rs189110490 Chr.9: 77797480-77797480 on Build GRCh38
rs776716957 Chr.9: 77797491-77797491 on Build GRCh38

View Full Product Details