Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 148807557-148807731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EZH2
Entrez Gene ID
2146
Gene Name
enhancer of zeste 2 polycomb repressive complex 2 subunit
Gene Aliases
ENX-1, ENX1, EZH1, EZH2b, KMT6, KMT6A, WVS, WVS2
UniGene
Hs.444082
Gene Chromosome Location
Chr.7: 148807372-148884662 on Build GRCh38
Transcripts
NM_001203247.1 NM_001203248.1 NM_001203249.1 NM_004456.4 NM_152998.2 XM_005249962.4 XM_005249963.4 XM_005249964.4 XM_011515883.2 XM_011515884.2 XM_011515885.2 XM_011515886.2 XM_011515887.2 XM_011515888.2 XM_011515889.2 XM_011515890.2 XM_011515891.2 XM_011515892.2 XM_011515893.2 XM_011515894.2 XM_011515895.2 XM_011515896.2 XM_011515897.2 XM_011515898.2 XM_017011817.1 XM_017011818.1 XM_017011819.1 XM_017011820.1 XM_017011821.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6950963 c.2210_2227del18 p.D737_V742delDALKYV Chr.7: 148807675-148807692 on Build GRCh38
144185 c.2226_2227insAAAACC p.V742_G743insKT Chr.7: 148807675-148807676 on Build GRCh38
5487424 c.2231_2232insA p.E745fs*>8 Chr.7: 148807670-148807671 on Build GRCh38
1315857 c.2232_2233insATC p.I744_E745insI Chr.7: 148807669-148807670 on Build GRCh38
5879069 c.2231_2232insGGCAT p.I744fs*>10 Chr.7: 148807670-148807671 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779629814 Chr.7: 148807652-148807652 on Build GRCh38
rs763362563 Chr.7: 148807626-148807626 on Build GRCh38
rs752038271 Chr.7: 148807627-148807627 on Build GRCh38
rs745948888 Chr.7: 148807639-148807639 on Build GRCh38
rs776312600 Chr.7: 148807703-148807703 on Build GRCh38
rs754610785 Chr.7: 148807643-148807644 on Build GRCh38
rs373732420 Chr.7: 148807630-148807630 on Build GRCh38
rs148678300 Chr.7: 148807589-148807589 on Build GRCh38
rs368565050 Chr.7: 148807631-148807631 on Build GRCh38
rs755736107 Chr.7: 148807643-148807643 on Build GRCh38
rs749969922 Chr.7: 148807642-148807642 on Build GRCh38
rs753561759 Chr.7: 148807624-148807625 on Build GRCh38
rs929618724 Chr.7: 148807614-148807614 on Build GRCh38
rs3217095 Chr.7: 148807625-148807625 on Build GRCh38
rs373975096 Chr.7: 148807670-148807670 on Build GRCh38
rs886990256 Chr.7: 148807691-148807691 on Build GRCh38
rs760581114 Chr.7: 148807622-148807622 on Build GRCh38
rs770439804 Chr.7: 148807640-148807640 on Build GRCh38
rs759825516 Chr.7: 148807620-148807620 on Build GRCh38
rs201597962 Chr.7: 148807625-148807625 on Build GRCh38
rs148228353 Chr.7: 148807676-148807676 on Build GRCh38
rs748102400 Chr.7: 148807679-148807679 on Build GRCh38
rs397515548 Chr.7: 148807669-148807669 on Build GRCh38
rs765595441 Chr.7: 148807622-148807622 on Build GRCh38
rs587783627 Chr.7: 148807666-148807666 on Build GRCh38
rs757722069 Chr.7: 148807633-148807633 on Build GRCh38

View Full Product Details